Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57546452-57546695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMTNL1
Entrez Gene ID
219537
Gene Name
smoothelin like 1
Gene Aliases
CHASM
UniGene
Hs.68756
Gene Chromosome Location
Chr.11: 57536841-57550274 on Build GRCh38
Transcripts
NM_001105565.2 XM_005273819.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
429292 c.1289C>A p.A430D Chr.11: 57546547-57546547 on Build GRCh38
6044857 c.1358G>A p.R453H Chr.11: 57546616-57546616 on Build GRCh38
3449745 c.1280G>A p.S427N Chr.11: 57546538-57546538 on Build GRCh38
4390766 c.1383C>T p.F461F Chr.11: 57546641-57546641 on Build GRCh38
1354783 c.1260_1262delCTT p.F421delF Chr.11: 57546518-57546520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778479649 Chr.11: 57546656-57546656 on Build GRCh38
rs564909263 Chr.11: 57546542-57546542 on Build GRCh38
rs931766318 Chr.11: 57546494-57546494 on Build GRCh38
rs909050893 Chr.11: 57546614-57546614 on Build GRCh38
rs751781075 Chr.11: 57546618-57546618 on Build GRCh38
rs758058873 Chr.11: 57546475-57546475 on Build GRCh38
rs770508595 Chr.11: 57546498-57546498 on Build GRCh38
rs540776815 Chr.11: 57546606-57546606 on Build GRCh38
rs372869577 Chr.11: 57546560-57546560 on Build GRCh38
rs756063656 Chr.11: 57546635-57546635 on Build GRCh38
rs372676547 Chr.11: 57546483-57546483 on Build GRCh38
rs763115924 Chr.11: 57546611-57546611 on Build GRCh38
rs755967024 Chr.11: 57546582-57546582 on Build GRCh38
rs570413222 Chr.11: 57546593-57546593 on Build GRCh38
rs761087610 Chr.11: 57546508-57546508 on Build GRCh38
rs777165740 Chr.11: 57546530-57546530 on Build GRCh38
rs769111420 Chr.11: 57546524-57546524 on Build GRCh38
rs748742577 Chr.11: 57546589-57546589 on Build GRCh38
rs765437133 Chr.11: 57546550-57546550 on Build GRCh38
rs370016574 Chr.11: 57546532-57546532 on Build GRCh38
rs903901084 Chr.11: 57546507-57546507 on Build GRCh38
rs767379074 Chr.11: 57546623-57546623 on Build GRCh38
rs747375658 Chr.11: 57546595-57546595 on Build GRCh38
rs756731519 Chr.11: 57546654-57546654 on Build GRCh38
rs781037832 Chr.11: 57546581-57546581 on Build GRCh38
rs754768636 Chr.11: 57546566-57546566 on Build GRCh38
rs543726004 Chr.11: 57546506-57546506 on Build GRCh38
rs547961415 Chr.11: 57546667-57546667 on Build GRCh38
rs1008457746 Chr.11: 57546526-57546526 on Build GRCh38
rs866660047 Chr.11: 57546576-57546576 on Build GRCh38
rs750460923 Chr.11: 57546554-57546554 on Build GRCh38
rs766745806 Chr.11: 57546616-57546616 on Build GRCh38
rs1049871544 Chr.11: 57546504-57546504 on Build GRCh38
rs778534246 Chr.11: 57546594-57546594 on Build GRCh38
rs559016337 Chr.11: 57546609-57546609 on Build GRCh38
rs748975609 Chr.11: 57546491-57546491 on Build GRCh38
rs762386856 Chr.11: 57546608-57546608 on Build GRCh38
rs762177270 Chr.11: 57546533-57546533 on Build GRCh38
rs776818800 Chr.11: 57546607-57546607 on Build GRCh38
rs921683114 Chr.11: 57546486-57546486 on Build GRCh38
rs369245699 Chr.11: 57546636-57546636 on Build GRCh38
rs200371755 Chr.11: 57546628-57546628 on Build GRCh38
rs947048820 Chr.11: 57546481-57546481 on Build GRCh38
rs377127608 Chr.11: 57546619-57546619 on Build GRCh38
rs777274265 Chr.11: 57546583-57546583 on Build GRCh38

View Full Product Details