Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103131504-103131775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFAIP2
Entrez Gene ID
7127
Gene Name
TNF alpha induced protein 2
Gene Aliases
B94, EXOC3L3
UniGene
Hs.525607
Gene Chromosome Location
Chr.14: 103121175-103137439 on Build GRCh38
Transcripts
NM_006291.2 XM_006720243.3 XM_011537112.2 XM_011537113.2 XM_011537114.2 XM_017021616.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4049256 c.1391C>T p.T464I Chr.14: 103131731-103131731 on Build GRCh38
5656458 c.1393C>G p.L465V Chr.14: 103131733-103131733 on Build GRCh38
1722812 c.1327C>T p.P443S Chr.14: 103131667-103131667 on Build GRCh38
6804915 c.1355G>C p.G452A Chr.14: 103131695-103131695 on Build GRCh38
4049257 c.1410T>C p.H470H Chr.14: 103131750-103131750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768759079 Chr.14: 103131601-103131601 on Build GRCh38
rs200163502 Chr.14: 103131682-103131682 on Build GRCh38
rs755395837 Chr.14: 103131651-103131651 on Build GRCh38
rs1028183707 Chr.14: 103131570-103131570 on Build GRCh38
rs986162971 Chr.14: 103131575-103131575 on Build GRCh38
rs750798325 Chr.14: 103131628-103131628 on Build GRCh38
rs61706103 Chr.14: 103131684-103131684 on Build GRCh38
rs778520719 Chr.14: 103131652-103131652 on Build GRCh38
rs947344239 Chr.14: 103131636-103131636 on Build GRCh38
rs868220171 Chr.14: 103131654-103131654 on Build GRCh38
rs752084520 Chr.14: 103131657-103131657 on Build GRCh38
rs762125363 Chr.14: 103131617-103131617 on Build GRCh38
rs544018632 Chr.14: 103131582-103131582 on Build GRCh38
rs761820807 Chr.14: 103131706-103131706 on Build GRCh38
rs777555606 Chr.14: 103131667-103131667 on Build GRCh38
rs564019638 Chr.14: 103131720-103131720 on Build GRCh38
rs765070275 Chr.14: 103131715-103131715 on Build GRCh38
rs952159787 Chr.14: 103131573-103131573 on Build GRCh38
rs758140553 Chr.14: 103131665-103131665 on Build GRCh38
rs761694831 Chr.14: 103131688-103131690 on Build GRCh38
rs561910000 Chr.14: 103131646-103131646 on Build GRCh38
rs773414077 Chr.14: 103131710-103131710 on Build GRCh38
rs760096089 Chr.14: 103131731-103131731 on Build GRCh38
rs761064513 Chr.14: 103131711-103131711 on Build GRCh38
rs745839940 Chr.14: 103131685-103131685 on Build GRCh38
rs572696113 Chr.14: 103131669-103131669 on Build GRCh38
rs772336180 Chr.14: 103131709-103131709 on Build GRCh38
rs767657930 Chr.14: 103131621-103131621 on Build GRCh38
rs2297064 Chr.14: 103131608-103131608 on Build GRCh38
rs976347714 Chr.14: 103131624-103131625 on Build GRCh38
rs761011435 Chr.14: 103131641-103131641 on Build GRCh38
rs762983370 Chr.14: 103131589-103131589 on Build GRCh38
rs769575979 Chr.14: 103131692-103131692 on Build GRCh38
rs766934516 Chr.14: 103131642-103131642 on Build GRCh38
rs765608185 Chr.14: 103131732-103131732 on Build GRCh38
rs774492555 Chr.14: 103131697-103131697 on Build GRCh38
rs140309001 Chr.14: 103131721-103131721 on Build GRCh38
rs576304605 Chr.14: 103131545-103131545 on Build GRCh38
rs995323673 Chr.14: 103131556-103131556 on Build GRCh38
rs1042945347 Chr.14: 103131649-103131649 on Build GRCh38
rs1013182517 Chr.14: 103131738-103131738 on Build GRCh38

View Full Product Details