Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39105569-39105822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAYSD1
Entrez Gene ID
55776
Gene Name
SAYSVFN motif domain containing 1
Gene Aliases
C6orf64
UniGene
Hs.58382
Gene Chromosome Location
Chr.6: 39104063-39115186 on Build GRCh38
Transcripts
NM_001304793.1 NM_018322.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5518665 c.406G>A p.V136I Chr.6: 39105578-39105578 on Build GRCh38
1198962 c.347T>C p.F116S Chr.6: 39105637-39105637 on Build GRCh38
4492219 c.393C>T p.F131F Chr.6: 39105591-39105591 on Build GRCh38
6547216 c.249G>A p.W83* Chr.6: 39105735-39105735 on Build GRCh38
742021 c.393C>G p.F131L Chr.6: 39105591-39105591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905416613 Chr.6: 39105757-39105757 on Build GRCh38
rs142879633 Chr.6: 39105589-39105589 on Build GRCh38
rs778290735 Chr.6: 39105677-39105677 on Build GRCh38
rs111858504 Chr.6: 39105645-39105645 on Build GRCh38
rs377248635 Chr.6: 39105711-39105711 on Build GRCh38
rs139146544 Chr.6: 39105732-39105732 on Build GRCh38
rs779278213 Chr.6: 39105646-39105646 on Build GRCh38
rs114633981 Chr.6: 39105771-39105771 on Build GRCh38
rs761871373 Chr.6: 39105719-39105719 on Build GRCh38
rs767067947 Chr.6: 39105720-39105720 on Build GRCh38
rs759750688 Chr.6: 39105705-39105705 on Build GRCh38
rs755773850 Chr.6: 39105754-39105754 on Build GRCh38
rs199703500 Chr.6: 39105772-39105772 on Build GRCh38
rs929034932 Chr.6: 39105643-39105643 on Build GRCh38
rs773008995 Chr.6: 39105720-39105724 on Build GRCh38
rs1047261961 Chr.6: 39105712-39105712 on Build GRCh38
rs562214094 Chr.6: 39105688-39105688 on Build GRCh38
rs11558532 Chr.6: 39105603-39105603 on Build GRCh38
rs751084796 Chr.6: 39105600-39105600 on Build GRCh38
rs923047858 Chr.6: 39105641-39105641 on Build GRCh38
rs756877962 Chr.6: 39105601-39105601 on Build GRCh38
rs752280318 Chr.6: 39105715-39105715 on Build GRCh38
rs760033844 Chr.6: 39105726-39105757 on Build GRCh38
rs112205512 Chr.6: 39105776-39105776 on Build GRCh38
rs749914403 Chr.6: 39105737-39105737 on Build GRCh38
rs769971435 Chr.6: 39105708-39105708 on Build GRCh38
rs528183094 Chr.6: 39105670-39105670 on Build GRCh38
rs960233610 Chr.6: 39105595-39105595 on Build GRCh38
rs776699499 Chr.6: 39105697-39105697 on Build GRCh38
rs764194841 Chr.6: 39105597-39105597 on Build GRCh38
rs147416208 Chr.6: 39105714-39105714 on Build GRCh38
rs748516328 Chr.6: 39105665-39105665 on Build GRCh38
rs780870017 Chr.6: 39105610-39105610 on Build GRCh38
rs755726898 Chr.6: 39105634-39105634 on Build GRCh38
rs750050425 Chr.6: 39105619-39105619 on Build GRCh38
rs548807253 Chr.6: 39105685-39105685 on Build GRCh38
rs111822360 Chr.6: 39105717-39105717 on Build GRCh38
rs941924869 Chr.6: 39105787-39105787 on Build GRCh38

View Full Product Details