Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45377061-45377322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYS1-DBNDD2
Entrez Gene ID
767557
Gene Name
SYS1-DBNDD2 readthrough (NMD candidate)
Gene Aliases
C20orf169-DBNDD2
UniGene
Hs.730643
Gene Chromosome Location
Chr.20: 45363169-45410610 on Build GRCh38
Transcripts
NR_003189.2 NM_014477.2 XM_011528790.2

Gene Symbol
TP53TG5
Entrez Gene ID
27296
Gene Name
TP53 target 5
Gene Aliases
C20orf10, CLG01
UniGene
Hs.710271
Gene Chromosome Location
Chr.20: 45373880-45378550 on Build GRCh38
Transcripts
NR_003189.2 NM_014477.2 XM_011528790.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3547015 c.147G>A p.L49L Chr.20: 45377319-45377319 on Build GRCh38
4098831 c.184C>A p.L62M Chr.20: 45377282-45377282 on Build GRCh38
7102020 c.174G>A p.R58R Chr.20: 45377292-45377292 on Build GRCh38
3841114 c.238C>T p.L80F Chr.20: 45377228-45377228 on Build GRCh38
6487046 c.242G>A p.R81H Chr.20: 45377224-45377224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762897962 Chr.20: 45377173-45377173 on Build GRCh38
rs948056801 Chr.20: 45377156-45377156 on Build GRCh38
rs751426346 Chr.20: 45377182-45377182 on Build GRCh38
rs2741651 Chr.20: 45377136-45377136 on Build GRCh38
rs147911582 Chr.20: 45377293-45377293 on Build GRCh38
rs754758610 Chr.20: 45377184-45377184 on Build GRCh38
rs917935424 Chr.20: 45377128-45377128 on Build GRCh38
rs373282656 Chr.20: 45377289-45377289 on Build GRCh38
rs777738513 Chr.20: 45377225-45377225 on Build GRCh38
rs370602916 Chr.20: 45377255-45377255 on Build GRCh38
rs767833115 Chr.20: 45377190-45377190 on Build GRCh38
rs772715528 Chr.20: 45377297-45377297 on Build GRCh38
rs2231616 Chr.20: 45377296-45377296 on Build GRCh38
rs183808821 Chr.20: 45377172-45377172 on Build GRCh38
rs758354293 Chr.20: 45377194-45377194 on Build GRCh38
rs1044134236 Chr.20: 45377214-45377214 on Build GRCh38
rs746292966 Chr.20: 45377262-45377262 on Build GRCh38
rs779558461 Chr.20: 45377261-45377261 on Build GRCh38
rs867134832 Chr.20: 45377270-45377270 on Build GRCh38
rs779918002 Chr.20: 45377224-45377224 on Build GRCh38
rs768952108 Chr.20: 45377127-45377127 on Build GRCh38
rs747784166 Chr.20: 45377295-45377295 on Build GRCh38
rs577494843 Chr.20: 45377282-45377283 on Build GRCh38
rs992244128 Chr.20: 45377098-45377098 on Build GRCh38
rs2231617 Chr.20: 45377174-45377174 on Build GRCh38
rs749324909 Chr.20: 45377254-45377254 on Build GRCh38
rs752830982 Chr.20: 45377205-45377205 on Build GRCh38
rs377199362 Chr.20: 45377226-45377226 on Build GRCh38
rs758347225 Chr.20: 45377257-45377258 on Build GRCh38

View Full Product Details