Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76306813-76307038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM47E-STBD1
Entrez Gene ID
100631383
Gene Name
FAM47E-STBD1 readthrough
Gene Aliases
-
UniGene
Hs.720509
Gene Chromosome Location
Chr.4: 76251700-76311130 on Build GRCh38
Transcripts
NM_001242939.1 NM_003943.4

Gene Symbol
STBD1
Entrez Gene ID
8987
Gene Name
starch binding domain 1
Gene Aliases
GENEX3414, GENX-3414
UniGene
Hs.109590
Gene Chromosome Location
Chr.4: 76306026-76311130 on Build GRCh38
Transcripts
NM_001242939.1 NM_003943.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1057653 c.79C>T p.P27S Chr.4: 76306848-76306848 on Build GRCh38
6893699 c.133G>A p.G45R Chr.4: 76306902-76306902 on Build GRCh38
4656407 c.220G>A p.E74K Chr.4: 76306989-76306989 on Build GRCh38
6384685 c.94A>G p.K32E Chr.4: 76306863-76306863 on Build GRCh38
7078788 c.103G>A p.D35N Chr.4: 76306872-76306872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773503088 Chr.4: 76306894-76306894 on Build GRCh38
rs371699339 Chr.4: 76306872-76306872 on Build GRCh38
rs147138225 Chr.4: 76306890-76306890 on Build GRCh38
rs372051485 Chr.4: 76306846-76306846 on Build GRCh38
rs780808206 Chr.4: 76306977-76306977 on Build GRCh38
rs755108417 Chr.4: 76306912-76306912 on Build GRCh38
rs776156534 Chr.4: 76306871-76306871 on Build GRCh38
rs761691334 Chr.4: 76306998-76306998 on Build GRCh38
rs202009814 Chr.4: 76306935-76306935 on Build GRCh38
rs768524371 Chr.4: 76306994-76306994 on Build GRCh38
rs775507048 Chr.4: 76306990-76306990 on Build GRCh38
rs34842707 Chr.4: 76306892-76306892 on Build GRCh38
rs114978333 Chr.4: 76306876-76306876 on Build GRCh38
rs758623598 Chr.4: 76306847-76306847 on Build GRCh38
rs151238403 Chr.4: 76306906-76306906 on Build GRCh38
rs775284657 Chr.4: 76306884-76306884 on Build GRCh38
rs774066686 Chr.4: 76306996-76306996 on Build GRCh38
rs766778477 Chr.4: 76307002-76307002 on Build GRCh38
rs747366979 Chr.4: 76306964-76306964 on Build GRCh38
rs1053464 Chr.4: 76306853-76306853 on Build GRCh38
rs368488147 Chr.4: 76306839-76306839 on Build GRCh38
rs756950678 Chr.4: 76306967-76306967 on Build GRCh38
rs766688719 Chr.4: 76306904-76306904 on Build GRCh38
rs758815009 Chr.4: 76306933-76306933 on Build GRCh38
rs765323665 Chr.4: 76306869-76306902 on Build GRCh38
rs368161631 Chr.4: 76306845-76306845 on Build GRCh38
rs750463429 Chr.4: 76306881-76306882 on Build GRCh38
rs766515008 Chr.4: 76306833-76306833 on Build GRCh38
rs769222855 Chr.4: 76306988-76306988 on Build GRCh38
rs765844033 Chr.4: 76306919-76306919 on Build GRCh38
rs759701839 Chr.4: 76307008-76307008 on Build GRCh38
rs777961792 Chr.4: 76306938-76306938 on Build GRCh38
rs753088427 Chr.4: 76306923-76306923 on Build GRCh38
rs745449050 Chr.4: 76306985-76306985 on Build GRCh38
rs755594376 Chr.4: 76306840-76306840 on Build GRCh38
rs777495733 Chr.4: 76306852-76306852 on Build GRCh38
rs199978693 Chr.4: 76306854-76306854 on Build GRCh38
rs1004763342 Chr.4: 76306898-76306898 on Build GRCh38
rs779552705 Chr.4: 76306842-76306842 on Build GRCh38
rs199535918 Chr.4: 76306903-76306903 on Build GRCh38

View Full Product Details