Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30580941-30581214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PES1
Entrez Gene ID
23481
Gene Name
pescadillo ribosomal biogenesis factor 1
Gene Aliases
PES
UniGene
Hs.517543
Gene Chromosome Location
Chr.22: 30576625-30607013 on Build GRCh38
Transcripts
NM_001243225.1 NM_001282327.1 NM_001282328.1 NM_014303.3 XM_006724173.1 XM_017028678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5859171 c.902C>T p.P301L Chr.22: 30581022-30581022 on Build GRCh38
5859170 c.911G>A p.G304E Chr.22: 30581013-30581013 on Build GRCh38
2937300 c.853C>T p.R285C Chr.22: 30581071-30581071 on Build GRCh38
7275275 c.825A>G p.K275K Chr.22: 30581099-30581099 on Build GRCh38
1220201 c.907G>A p.D303N Chr.22: 30581017-30581017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144358121 Chr.22: 30581014-30581014 on Build GRCh38
rs751574321 Chr.22: 30581039-30581039 on Build GRCh38
rs2108110 Chr.22: 30581027-30581027 on Build GRCh38
rs745626369 Chr.22: 30580968-30580968 on Build GRCh38
rs557841800 Chr.22: 30581073-30581073 on Build GRCh38
rs9608991 Chr.22: 30580986-30580986 on Build GRCh38
rs761411876 Chr.22: 30581004-30581004 on Build GRCh38
rs775660770 Chr.22: 30581135-30581135 on Build GRCh38
rs764547924 Chr.22: 30581139-30581139 on Build GRCh38
rs372779600 Chr.22: 30581071-30581071 on Build GRCh38
rs768410074 Chr.22: 30581086-30581086 on Build GRCh38
rs769503482 Chr.22: 30580969-30580969 on Build GRCh38
rs377464980 Chr.22: 30581117-30581117 on Build GRCh38
rs375893770 Chr.22: 30580966-30580966 on Build GRCh38
rs967680677 Chr.22: 30581054-30581054 on Build GRCh38
rs750609057 Chr.22: 30581069-30581069 on Build GRCh38
rs556088204 Chr.22: 30581019-30581019 on Build GRCh38
rs373902653 Chr.22: 30581138-30581138 on Build GRCh38
rs747611657 Chr.22: 30581094-30581094 on Build GRCh38
rs749119249 Chr.22: 30581075-30581075 on Build GRCh38
rs148346090 Chr.22: 30581018-30581018 on Build GRCh38
rs559943361 Chr.22: 30581145-30581145 on Build GRCh38
rs1014551834 Chr.22: 30581063-30581063 on Build GRCh38
rs752087745 Chr.22: 30581140-30581140 on Build GRCh38
rs979338334 Chr.22: 30581056-30581056 on Build GRCh38
rs933979516 Chr.22: 30581182-30581182 on Build GRCh38
rs775392337 Chr.22: 30580972-30580972 on Build GRCh38
rs770677363 Chr.22: 30581126-30581126 on Build GRCh38
rs763383915 Chr.22: 30581137-30581137 on Build GRCh38
rs369961765 Chr.22: 30580980-30580980 on Build GRCh38
rs575214281 Chr.22: 30581070-30581070 on Build GRCh38
rs779023930 Chr.22: 30581091-30581091 on Build GRCh38
rs112894042 Chr.22: 30581196-30581196 on Build GRCh38
rs757416031 Chr.22: 30580965-30580965 on Build GRCh38
rs773937656 Chr.22: 30581003-30581003 on Build GRCh38
rs979357953 Chr.22: 30581103-30581103 on Build GRCh38
rs758819706 Chr.22: 30581038-30581038 on Build GRCh38
rs201919075 Chr.22: 30581144-30581144 on Build GRCh38
rs368763687 Chr.22: 30581118-30581118 on Build GRCh38
rs768693936 Chr.22: 30581001-30581001 on Build GRCh38
rs773019625 Chr.22: 30581017-30581017 on Build GRCh38
rs944723115 Chr.22: 30581110-30581110 on Build GRCh38
rs781346992 Chr.22: 30581068-30581068 on Build GRCh38
rs757362887 Chr.22: 30581048-30581048 on Build GRCh38
rs762360776 Chr.22: 30581142-30581142 on Build GRCh38
rs760240387 Chr.22: 30581125-30581125 on Build GRCh38

View Full Product Details