Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6319218-6319467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCDBP
Entrez Gene ID
112464
Gene Name
protein kinase C delta binding protein
Gene Aliases
CAVIN3, HSRBC, SRBC, cavin-3
UniGene
Hs.434044
Gene Chromosome Location
Chr.11: 6318946-6320510 on Build GRCh38
Transcripts
NM_145040.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1475682 c.720C>T p.T240T Chr.11: 6319229-6319229 on Build GRCh38
4717721 c.618G>A p.P206P Chr.11: 6319331-6319331 on Build GRCh38
6755804 c.596C>T p.P199L Chr.11: 6319353-6319353 on Build GRCh38
267260 c.565C>A p.L189I Chr.11: 6319384-6319384 on Build GRCh38
4717722 c.539G>A p.R180H Chr.11: 6319410-6319410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751400273 Chr.11: 6319384-6319384 on Build GRCh38
rs369154864 Chr.11: 6319290-6319290 on Build GRCh38
rs141194842 Chr.11: 6319381-6319381 on Build GRCh38
rs12570 Chr.11: 6319295-6319295 on Build GRCh38
rs543801330 Chr.11: 6319323-6319323 on Build GRCh38
rs143254060 Chr.11: 6319392-6319392 on Build GRCh38
rs767287993 Chr.11: 6319386-6319386 on Build GRCh38
rs780849905 Chr.11: 6319295-6319296 on Build GRCh38
rs777880839 Chr.11: 6319396-6319396 on Build GRCh38
rs371263806 Chr.11: 6319348-6319348 on Build GRCh38
rs771036928 Chr.11: 6319334-6319334 on Build GRCh38
rs774548548 Chr.11: 6319267-6319267 on Build GRCh38
rs751555322 Chr.11: 6319271-6319272 on Build GRCh38
rs555002736 Chr.11: 6319296-6319296 on Build GRCh38
rs927386218 Chr.11: 6319285-6319285 on Build GRCh38
rs139734641 Chr.11: 6319311-6319311 on Build GRCh38
rs763059366 Chr.11: 6319248-6319259 on Build GRCh38
rs781065717 Chr.11: 6319322-6319322 on Build GRCh38
rs1027592603 Chr.11: 6319448-6319448 on Build GRCh38
rs747554370 Chr.11: 6319444-6319444 on Build GRCh38
rs150739333 Chr.11: 6319260-6319260 on Build GRCh38
rs749282736 Chr.11: 6319406-6319406 on Build GRCh38
rs757038346 Chr.11: 6319280-6319281 on Build GRCh38
rs763997486 Chr.11: 6319264-6319264 on Build GRCh38
rs760142956 Chr.11: 6319268-6319268 on Build GRCh38
rs772641029 Chr.11: 6319352-6319352 on Build GRCh38
rs778937529 Chr.11: 6319418-6319418 on Build GRCh38
rs1001980082 Chr.11: 6319426-6319426 on Build GRCh38
rs139113360 Chr.11: 6319266-6319266 on Build GRCh38
rs983941609 Chr.11: 6319288-6319288 on Build GRCh38
rs781381437 Chr.11: 6319249-6319249 on Build GRCh38
rs144522181 Chr.11: 6319337-6319337 on Build GRCh38
rs754661198 Chr.11: 6319385-6319385 on Build GRCh38
rs932277589 Chr.11: 6319331-6319331 on Build GRCh38
rs917494370 Chr.11: 6319299-6319299 on Build GRCh38
rs773366841 Chr.11: 6319259-6319259 on Build GRCh38
rs764992045 Chr.11: 6319291-6319291 on Build GRCh38
rs769260829 Chr.11: 6319447-6319447 on Build GRCh38
rs373445694 Chr.11: 6319332-6319332 on Build GRCh38
rs146538294 Chr.11: 6319353-6319353 on Build GRCh38
rs746260843 Chr.11: 6319424-6319424 on Build GRCh38
rs970440456 Chr.11: 6319240-6319240 on Build GRCh38
rs531215074 Chr.11: 6319394-6319394 on Build GRCh38
rs764904358 Chr.11: 6319377-6319377 on Build GRCh38
rs748174686 Chr.11: 6319250-6319250 on Build GRCh38
rs149823183 Chr.11: 6319271-6319271 on Build GRCh38
rs201987135 Chr.11: 6319246-6319246 on Build GRCh38
rs961288525 Chr.11: 6319298-6319298 on Build GRCh38
rs371948974 Chr.11: 6319258-6319258 on Build GRCh38
rs1028188353 Chr.11: 6319269-6319269 on Build GRCh38
rs773615461 Chr.11: 6319346-6319346 on Build GRCh38
rs578200005 Chr.11: 6319330-6319330 on Build GRCh38
rs776315629 Chr.11: 6319275-6319275 on Build GRCh38
rs757534658 Chr.11: 6319410-6319410 on Build GRCh38
rs765898204 Chr.11: 6319383-6319383 on Build GRCh38
rs750032041 Chr.11: 6319330-6319330 on Build GRCh38
rs376757809 Chr.11: 6319380-6319380 on Build GRCh38
rs538451811 Chr.11: 6319264-6319264 on Build GRCh38
rs768446790 Chr.11: 6319242-6319242 on Build GRCh38
rs996001883 Chr.11: 6319388-6319388 on Build GRCh38
rs772620232 Chr.11: 6319427-6319427 on Build GRCh38
rs761247913 Chr.11: 6319367-6319367 on Build GRCh38

View Full Product Details