Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28491710-28491935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC13A2
Entrez Gene ID
9058
Gene Name
solute carrier family 13 member 2
Gene Aliases
NADC1, NaCT, NaDC-1, SDCT1
UniGene
Hs.102307
Gene Chromosome Location
Chr.17: 28473358-28497781 on Build GRCh38
Transcripts
NM_001145975.1 NM_003984.3 NR_027384.1 XM_006722165.2 XM_011525450.2 XM_011525451.1 XM_011525452.1 XM_011525453.2 XM_011525454.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1479363 c.774C>A p.G258G Chr.17: 28491748-28491748 on Build GRCh38
1588746 c.924C>T p.N308N Chr.17: 28491751-28491751 on Build GRCh38
7194210 c.847T>C p.L283L Chr.17: 28491821-28491821 on Build GRCh38
976878 c.777C>T p.N259N Chr.17: 28491751-28491751 on Build GRCh38
4064909 c.800T>A p.F267Y Chr.17: 28491774-28491774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782061711 Chr.17: 28491884-28491885 on Build GRCh38
rs781903288 Chr.17: 28491873-28491873 on Build GRCh38
rs782799413 Chr.17: 28491814-28491814 on Build GRCh38
rs1010601561 Chr.17: 28491761-28491761 on Build GRCh38
rs781815137 Chr.17: 28491818-28491818 on Build GRCh38
rs368817537 Chr.17: 28491869-28491869 on Build GRCh38
rs781821669 Chr.17: 28491764-28491764 on Build GRCh38
rs782584247 Chr.17: 28491884-28491884 on Build GRCh38
rs974133679 Chr.17: 28491907-28491907 on Build GRCh38
rs782472673 Chr.17: 28491746-28491746 on Build GRCh38
rs782338452 Chr.17: 28491893-28491893 on Build GRCh38
rs960875983 Chr.17: 28491819-28491819 on Build GRCh38
rs782543241 Chr.17: 28491740-28491740 on Build GRCh38
rs782175078 Chr.17: 28491795-28491795 on Build GRCh38
rs117154964 Chr.17: 28491745-28491745 on Build GRCh38
rs79182027 Chr.17: 28491830-28491830 on Build GRCh38
rs782230907 Chr.17: 28491890-28491890 on Build GRCh38
rs782622258 Chr.17: 28491870-28491870 on Build GRCh38
rs991289455 Chr.17: 28491853-28491853 on Build GRCh38
rs916691918 Chr.17: 28491866-28491866 on Build GRCh38
rs781992542 Chr.17: 28491792-28491792 on Build GRCh38
rs782603070 Chr.17: 28491747-28491747 on Build GRCh38
rs890293411 Chr.17: 28491738-28491738 on Build GRCh38
rs782742301 Chr.17: 28491752-28491752 on Build GRCh38
rs11568461 Chr.17: 28491734-28491734 on Build GRCh38
rs112638123 Chr.17: 28491800-28491800 on Build GRCh38
rs145196579 Chr.17: 28491763-28491763 on Build GRCh38
rs782082795 Chr.17: 28491769-28491769 on Build GRCh38
rs375491240 Chr.17: 28491766-28491766 on Build GRCh38
rs1009027866 Chr.17: 28491739-28491739 on Build GRCh38
rs782768888 Chr.17: 28491736-28491736 on Build GRCh38
rs782772161 Chr.17: 28491782-28491782 on Build GRCh38
rs782756833 Chr.17: 28491852-28491852 on Build GRCh38
rs978315560 Chr.17: 28491794-28491794 on Build GRCh38
rs781817670 Chr.17: 28491865-28491865 on Build GRCh38
rs782047559 Chr.17: 28491842-28491842 on Build GRCh38
rs782534929 Chr.17: 28491883-28491883 on Build GRCh38

View Full Product Details