Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85848044-85848297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST3GAL5
Entrez Gene ID
8869
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 5
Gene Aliases
SATI, SIAT9, SIATGM3S, ST3GalV
UniGene
Hs.415117
Gene Chromosome Location
Chr.2: 85839148-85889034 on Build GRCh38
Transcripts
NM_001042437.1 NM_003896.3 XM_005264630.4 XM_011533143.2 XM_017005199.1 XM_017005200.1 XM_017005201.1 XM_017005202.1 XM_017005203.1 XM_017005204.1 XM_017005205.1 XM_017005206.1 XM_017005207.1 XM_017005208.1 XM_017005209.1 XM_017005210.1 XM_017005211.1 XM_017005212.1 XM_017005213.1 XM_017005214.1 XM_017005215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6791079 c.361C>T p.R121C Chr.2: 85848162-85848162 on Build GRCh38
4980552 c.300G>T p.K100N Chr.2: 85848154-85848154 on Build GRCh38
4918226 c.321G>A p.A107A Chr.2: 85848133-85848133 on Build GRCh38
4096076 c.345C>T p.S115S Chr.2: 85848109-85848109 on Build GRCh38
1409807 c.320C>T p.A107V Chr.2: 85848134-85848134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745590126 Chr.2: 85848080-85848080 on Build GRCh38
rs768713355 Chr.2: 85848180-85848180 on Build GRCh38
rs773350108 Chr.2: 85848189-85848189 on Build GRCh38
rs368983131 Chr.2: 85848167-85848167 on Build GRCh38
rs907536067 Chr.2: 85848144-85848144 on Build GRCh38
rs775825046 Chr.2: 85848246-85848246 on Build GRCh38
rs368878731 Chr.2: 85848109-85848109 on Build GRCh38
rs771210773 Chr.2: 85848192-85848192 on Build GRCh38
rs766447858 Chr.2: 85848110-85848110 on Build GRCh38
rs895373016 Chr.2: 85848259-85848259 on Build GRCh38
rs754643632 Chr.2: 85848170-85848170 on Build GRCh38
rs1055331435 Chr.2: 85848258-85848258 on Build GRCh38
rs753248662 Chr.2: 85848123-85848123 on Build GRCh38
rs373566687 Chr.2: 85848211-85848211 on Build GRCh38
rs779973935 Chr.2: 85848162-85848162 on Build GRCh38
rs759852193 Chr.2: 85848118-85848118 on Build GRCh38
rs867142071 Chr.2: 85848263-85848263 on Build GRCh38
rs777069591 Chr.2: 85848117-85848117 on Build GRCh38
rs144154879 Chr.2: 85848161-85848161 on Build GRCh38
rs144270260 Chr.2: 85848133-85848133 on Build GRCh38
rs772327731 Chr.2: 85848101-85848101 on Build GRCh38
rs374363945 Chr.2: 85848138-85848138 on Build GRCh38
rs368298013 Chr.2: 85848190-85848190 on Build GRCh38
rs762954685 Chr.2: 85848099-85848099 on Build GRCh38
rs770485013 Chr.2: 85848066-85848066 on Build GRCh38
rs140019292 Chr.2: 85848134-85848134 on Build GRCh38
rs1003757561 Chr.2: 85848227-85848227 on Build GRCh38
rs148420220 Chr.2: 85848088-85848088 on Build GRCh38
rs747005799 Chr.2: 85848191-85848191 on Build GRCh38
rs553252258 Chr.2: 85848212-85848212 on Build GRCh38
rs775530516 Chr.2: 85848097-85848097 on Build GRCh38
rs940418821 Chr.2: 85848155-85848155 on Build GRCh38
rs376353273 Chr.2: 85848249-85848249 on Build GRCh38
rs920382201 Chr.2: 85848226-85848226 on Build GRCh38
rs377046345 Chr.2: 85848108-85848108 on Build GRCh38
rs765551315 Chr.2: 85848119-85848119 on Build GRCh38
rs756946690 Chr.2: 85848145-85848145 on Build GRCh38
rs781044723 Chr.2: 85848075-85848075 on Build GRCh38
rs978587485 Chr.2: 85848095-85848095 on Build GRCh38
rs780658012 Chr.2: 85848146-85848146 on Build GRCh38
rs765643288 Chr.2: 85848224-85848224 on Build GRCh38

View Full Product Details