Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39513473-39513746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIEF1
Entrez Gene ID
54471
Gene Name
mitochondrial elongation factor 1
Gene Aliases
HSU79252, MID51, SMCR7L, dJ1104E15.3
UniGene
Hs.728085
Gene Chromosome Location
Chr.22: 39500100-39518134 on Build GRCh38
Transcripts
NM_001304564.1 NM_019008.5 NR_130789.1 NR_130790.1 XM_011530236.1 XM_011530237.1 XM_011530238.1 XM_017028838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5588984 c.712C>T p.P238S Chr.22: 39513643-39513643 on Build GRCh38
478980 c.749G>A p.R250H Chr.22: 39513680-39513680 on Build GRCh38
7161515 c.666C>T p.D222D Chr.22: 39513597-39513597 on Build GRCh38
7241299 c.685G>T p.G229C Chr.22: 39513616-39513616 on Build GRCh38
6400691 c.700C>A p.R234S Chr.22: 39513631-39513631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774154590 Chr.22: 39513620-39513620 on Build GRCh38
rs751601277 Chr.22: 39513507-39513507 on Build GRCh38
rs781546827 Chr.22: 39513531-39513531 on Build GRCh38
rs759722178 Chr.22: 39513632-39513632 on Build GRCh38
rs909642836 Chr.22: 39513655-39513655 on Build GRCh38
rs772998230 Chr.22: 39513718-39513718 on Build GRCh38
rs200199866 Chr.22: 39513678-39513678 on Build GRCh38
rs778255699 Chr.22: 39513551-39513551 on Build GRCh38
rs757705982 Chr.22: 39513654-39513654 on Build GRCh38
rs138446352 Chr.22: 39513595-39513595 on Build GRCh38
rs202201776 Chr.22: 39513609-39513609 on Build GRCh38
rs989590737 Chr.22: 39513576-39513576 on Build GRCh38
rs2232091 Chr.22: 39513721-39513721 on Build GRCh38
rs767475762 Chr.22: 39513634-39513634 on Build GRCh38
rs747820697 Chr.22: 39513713-39513713 on Build GRCh38
rs753012187 Chr.22: 39513636-39513636 on Build GRCh38
rs879462424 Chr.22: 39513720-39513721 on Build GRCh38
rs762892107 Chr.22: 39513605-39513605 on Build GRCh38
rs757786808 Chr.22: 39513570-39513570 on Build GRCh38
rs779538701 Chr.22: 39513656-39513656 on Build GRCh38
rs776401340 Chr.22: 39513581-39513581 on Build GRCh38
rs367952485 Chr.22: 39513500-39513500 on Build GRCh38
rs766392664 Chr.22: 39513502-39513502 on Build GRCh38
rs756360670 Chr.22: 39513646-39513646 on Build GRCh38
rs763011873 Chr.22: 39513545-39513545 on Build GRCh38
rs746345828 Chr.22: 39513666-39513666 on Build GRCh38
rs746460028 Chr.22: 39513573-39513573 on Build GRCh38
rs199993786 Chr.22: 39513535-39513535 on Build GRCh38
rs2232090 Chr.22: 39513717-39513717 on Build GRCh38
rs770621693 Chr.22: 39513723-39513723 on Build GRCh38
rs768044495 Chr.22: 39513579-39513579 on Build GRCh38
rs754193765 Chr.22: 39513652-39513652 on Build GRCh38
rs201294596 Chr.22: 39513571-39513571 on Build GRCh38
rs756599818 Chr.22: 39513537-39513537 on Build GRCh38
rs374798824 Chr.22: 39513499-39513499 on Build GRCh38
rs755162749 Chr.22: 39513522-39513522 on Build GRCh38
rs780650405 Chr.22: 39513679-39513679 on Build GRCh38
rs764571206 Chr.22: 39513651-39513651 on Build GRCh38
rs916942251 Chr.22: 39513602-39513602 on Build GRCh38
rs979884380 Chr.22: 39513630-39513630 on Build GRCh38
rs749839535 Chr.22: 39513563-39513563 on Build GRCh38
rs200542861 Chr.22: 39513596-39513596 on Build GRCh38
rs772943231 Chr.22: 39513604-39513604 on Build GRCh38

View Full Product Details