Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19894287-19894560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUD3
Entrez Gene ID
23252
Gene Name
OTU deubiquitinase 3
Gene Aliases
DUBA4
UniGene
Hs.374987
Gene Chromosome Location
Chr.1: 19881711-19912945 on Build GRCh38
Transcripts
NM_015207.1 XM_005245792.3 XM_005245793.3 XM_005245794.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4229189 c.474T>G p.P158P Chr.1: 19894471-19894471 on Build GRCh38
6990792 c.460C>G p.Q154E Chr.1: 19894457-19894457 on Build GRCh38
4571179 c.398T>C p.F133S Chr.1: 19894395-19894395 on Build GRCh38
902303 c.431G>T p.R144I Chr.1: 19894428-19894428 on Build GRCh38
5845126 c.404G>A p.G135D Chr.1: 19894401-19894401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs386629259 Chr.1: 19894523-19894536 on Build GRCh38
rs778112027 Chr.1: 19894412-19894412 on Build GRCh38
rs775945726 Chr.1: 19894478-19894478 on Build GRCh38
rs1038708387 Chr.1: 19894356-19894356 on Build GRCh38
rs774317857 Chr.1: 19894500-19894500 on Build GRCh38
rs779266574 Chr.1: 19894441-19894441 on Build GRCh38
rs760872074 Chr.1: 19894514-19894514 on Build GRCh38
rs780283976 Chr.1: 19894523-19894531 on Build GRCh38
rs966283051 Chr.1: 19894389-19894389 on Build GRCh38
rs1018671567 Chr.1: 19894365-19894365 on Build GRCh38
rs752860823 Chr.1: 19894390-19894390 on Build GRCh38
rs369295464 Chr.1: 19894494-19894494 on Build GRCh38
rs373159791 Chr.1: 19894353-19894353 on Build GRCh38
rs759370523 Chr.1: 19894508-19894508 on Build GRCh38
rs375869805 Chr.1: 19894428-19894428 on Build GRCh38
rs769174843 Chr.1: 19894483-19894483 on Build GRCh38
rs772719708 Chr.1: 19894333-19894333 on Build GRCh38
rs747744486 Chr.1: 19894482-19894482 on Build GRCh38
rs772710655 Chr.1: 19894484-19894484 on Build GRCh38
rs199878632 Chr.1: 19894523-19894530 on Build GRCh38
rs756122812 Chr.1: 19894399-19894399 on Build GRCh38
rs1057341948 Chr.1: 19894486-19894486 on Build GRCh38
rs372457119 Chr.1: 19894518-19894518 on Build GRCh38
rs553747914 Chr.1: 19894340-19894340 on Build GRCh38
rs752561077 Chr.1: 19894511-19894511 on Build GRCh38
rs935697141 Chr.1: 19894460-19894460 on Build GRCh38
rs200581344 Chr.1: 19894533-19894533 on Build GRCh38
rs764983325 Chr.1: 19894332-19894332 on Build GRCh38
rs767519303 Chr.1: 19894384-19894384 on Build GRCh38
rs186432684 Chr.1: 19894536-19894536 on Build GRCh38
rs746185753 Chr.1: 19894466-19894466 on Build GRCh38
rs762871877 Chr.1: 19894339-19894339 on Build GRCh38
rs199563201 Chr.1: 19894536-19894537 on Build GRCh38
rs757447481 Chr.1: 19894523-19894523 on Build GRCh38
rs779067766 Chr.1: 19894525-19894525 on Build GRCh38
rs762524755 Chr.1: 19894490-19894490 on Build GRCh38
rs372964689 Chr.1: 19894521-19894529 on Build GRCh38
rs565425521 Chr.1: 19894463-19894463 on Build GRCh38
rs751342084 Chr.1: 19894349-19894349 on Build GRCh38
rs749470027 Chr.1: 19894423-19894423 on Build GRCh38
rs757436022 Chr.1: 19894424-19894424 on Build GRCh38
rs772599029 Chr.1: 19894467-19894467 on Build GRCh38
rs764088933 Chr.1: 19894517-19894517 on Build GRCh38
rs571921740 Chr.1: 19894492-19894492 on Build GRCh38

View Full Product Details