Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63915526-63915751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCOR2
Entrez Gene ID
283248
Gene Name
REST corepressor 2
Gene Aliases
-
UniGene
Hs.98788
Gene Chromosome Location
Chr.11: 63911221-63916844 on Build GRCh38
Transcripts
NM_173587.3 XM_005273932.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1355596 c.142G>A p.V48I Chr.11: 63915597-63915597 on Build GRCh38
429509 c.163G>T p.V55L Chr.11: 63915576-63915576 on Build GRCh38
6363006 c.163G>A p.V55I Chr.11: 63915576-63915576 on Build GRCh38
7426273 c.129C>G p.D43E Chr.11: 63915610-63915610 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749806083 Chr.11: 63915589-63915589 on Build GRCh38
rs767780951 Chr.11: 63915599-63915599 on Build GRCh38
rs755801705 Chr.11: 63915656-63915656 on Build GRCh38
rs747798336 Chr.11: 63915640-63915642 on Build GRCh38
rs370851174 Chr.11: 63915580-63915580 on Build GRCh38
rs370499522 Chr.11: 63915690-63915690 on Build GRCh38
rs1036022082 Chr.11: 63915635-63915635 on Build GRCh38
rs760785652 Chr.11: 63915617-63915617 on Build GRCh38
rs143904903 Chr.11: 63915576-63915576 on Build GRCh38
rs753043727 Chr.11: 63915650-63915650 on Build GRCh38
rs1010385785 Chr.11: 63915552-63915552 on Build GRCh38
rs533854869 Chr.11: 63915678-63915678 on Build GRCh38
rs562198771 Chr.11: 63915711-63915711 on Build GRCh38
rs762912990 Chr.11: 63915556-63915556 on Build GRCh38
rs145688255 Chr.11: 63915597-63915597 on Build GRCh38
rs577816771 Chr.11: 63915618-63915618 on Build GRCh38
rs748802692 Chr.11: 63915657-63915657 on Build GRCh38
rs1034054539 Chr.11: 63915609-63915609 on Build GRCh38
rs909112406 Chr.11: 63915685-63915685 on Build GRCh38
rs373012221 Chr.11: 63915547-63915547 on Build GRCh38
rs918671077 Chr.11: 63915714-63915714 on Build GRCh38
rs755497445 Chr.11: 63915640-63915640 on Build GRCh38
rs967300178 Chr.11: 63915686-63915686 on Build GRCh38
rs775363574 Chr.11: 63915548-63915548 on Build GRCh38
rs745554448 Chr.11: 63915653-63915653 on Build GRCh38
rs551168351 Chr.11: 63915659-63915659 on Build GRCh38
rs778747928 Chr.11: 63915622-63915622 on Build GRCh38
rs779356619 Chr.11: 63915647-63915647 on Build GRCh38
rs1012444249 Chr.11: 63915691-63915691 on Build GRCh38
rs531440547 Chr.11: 63915623-63915623 on Build GRCh38
rs768520668 Chr.11: 63915561-63915561 on Build GRCh38
rs1020153981 Chr.11: 63915683-63915683 on Build GRCh38
rs1020053570 Chr.11: 63915569-63915569 on Build GRCh38
rs750685721 Chr.11: 63915616-63915616 on Build GRCh38
rs200765894 Chr.11: 63915652-63915652 on Build GRCh38
rs961868918 Chr.11: 63915697-63915697 on Build GRCh38
rs1009189882 Chr.11: 63915677-63915677 on Build GRCh38
rs546557957 Chr.11: 63915721-63915721 on Build GRCh38
rs971835137 Chr.11: 63915707-63915707 on Build GRCh38
rs747736890 Chr.11: 63915577-63915577 on Build GRCh38
rs571159384 Chr.11: 63915674-63915674 on Build GRCh38

View Full Product Details