Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17504560-17504833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USH1C
Entrez Gene ID
10083
Gene Name
USH1 protein network component harmonin
Gene Aliases
AIE-75, DFNB18, DFNB18A, NY-CO-37, NY-CO-38, PDZ-45, PDZ-73, PDZ-73/NY-CO-38, PDZ73, PDZD7C, ush1cpst
UniGene
Hs.502072
Gene Chromosome Location
Chr.11: 17493895-17544416 on Build GRCh38
Transcripts
NM_001297764.1 NM_005709.3 NM_153676.3 NR_123738.1 XM_011519832.2 XM_017017072.1 XM_017017073.1 XM_017017074.1 XM_017017075.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1746161 c.2151G>A p.K717K Chr.11: 17504680-17504680 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs984759697 Chr.11: 17504588-17504588 on Build GRCh38
rs761925493 Chr.11: 17504640-17504640 on Build GRCh38
rs143005574 Chr.11: 17504604-17504604 on Build GRCh38
rs796278158 Chr.11: 17504708-17504709 on Build GRCh38
rs569714771 Chr.11: 17504682-17504682 on Build GRCh38
rs986503748 Chr.11: 17504718-17504720 on Build GRCh38
rs534537152 Chr.11: 17504744-17504744 on Build GRCh38
rs146451547 Chr.11: 17504664-17504664 on Build GRCh38
rs777109466 Chr.11: 17504601-17504601 on Build GRCh38
rs375917372 Chr.11: 17504742-17504742 on Build GRCh38
rs781721922 Chr.11: 17504724-17504724 on Build GRCh38
rs769086751 Chr.11: 17504722-17504722 on Build GRCh38
rs764059609 Chr.11: 17504676-17504676 on Build GRCh38
rs565783252 Chr.11: 17504656-17504656 on Build GRCh38
rs190638847 Chr.11: 17504774-17504774 on Build GRCh38
rs76769358 Chr.11: 17504709-17504709 on Build GRCh38
rs747358137 Chr.11: 17504715-17504715 on Build GRCh38
rs200386499 Chr.11: 17504708-17504708 on Build GRCh38
rs551491679 Chr.11: 17504675-17504675 on Build GRCh38
rs75810555 Chr.11: 17504626-17504626 on Build GRCh38
rs746677181 Chr.11: 17504797-17504797 on Build GRCh38
rs752664237 Chr.11: 17504673-17504673 on Build GRCh38
rs375498939 Chr.11: 17504717-17504717 on Build GRCh38
rs780514445 Chr.11: 17504710-17504710 on Build GRCh38
rs748807658 Chr.11: 17504603-17504603 on Build GRCh38
rs773470577 Chr.11: 17504655-17504655 on Build GRCh38
rs562391372 Chr.11: 17504631-17504631 on Build GRCh38
rs11827813 Chr.11: 17504600-17504600 on Build GRCh38
rs778438766 Chr.11: 17504697-17504697 on Build GRCh38
rs34335652 Chr.11: 17504708-17504709 on Build GRCh38
rs892303634 Chr.11: 17504812-17504812 on Build GRCh38
rs767006224 Chr.11: 17504707-17504708 on Build GRCh38
rs748238450 Chr.11: 17504728-17504728 on Build GRCh38
rs763123373 Chr.11: 17504734-17504734 on Build GRCh38
rs138547759 Chr.11: 17504635-17504635 on Build GRCh38
rs778622894 Chr.11: 17504613-17504613 on Build GRCh38
rs573844196 Chr.11: 17504735-17504735 on Build GRCh38
rs773312528 Chr.11: 17504731-17504731 on Build GRCh38
rs761194766 Chr.11: 17504708-17504708 on Build GRCh38
rs368026455 Chr.11: 17504619-17504619 on Build GRCh38
rs200889109 Chr.11: 17504637-17504637 on Build GRCh38
rs142322601 Chr.11: 17504652-17504652 on Build GRCh38
rs774027072 Chr.11: 17504706-17504707 on Build GRCh38
rs756777467 Chr.11: 17504695-17504695 on Build GRCh38
rs397848183 Chr.11: 17504719-17504720 on Build GRCh38
rs907337685 Chr.11: 17504755-17504755 on Build GRCh38
rs1052711735 Chr.11: 17504608-17504608 on Build GRCh38
rs993637154 Chr.11: 17504773-17504773 on Build GRCh38
rs769853983 Chr.11: 17504729-17504729 on Build GRCh38
rs7932625 Chr.11: 17504586-17504586 on Build GRCh38
rs1052389617 Chr.11: 17504803-17504803 on Build GRCh38
rs760457471 Chr.11: 17504736-17504736 on Build GRCh38

View Full Product Details