Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184852355-184852624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS8
Entrez Gene ID
23355
Gene Name
VPS8, CORVET complex subunit
Gene Aliases
KIAA0804
UniGene
Hs.269263
Gene Chromosome Location
Chr.3: 184812137-185052614 on Build GRCh38
Transcripts
NM_001009921.2 NM_015303.3 XM_005247251.3 XM_005247253.4 XM_006713556.3 XM_011512600.2 XM_011512601.2 XM_017006040.1 XM_017006041.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6820758 c.786C>T p.N262N Chr.3: 184852532-184852532 on Build GRCh38
6436322 c.783C>A p.C261* Chr.3: 184852529-184852529 on Build GRCh38
5669359 c.810A>G p.E270E Chr.3: 184852556-184852556 on Build GRCh38
1484923 c.766C>G p.P256A Chr.3: 184852512-184852512 on Build GRCh38
4115830 c.787G>A p.D263N Chr.3: 184852533-184852533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754322988 Chr.3: 184852581-184852581 on Build GRCh38
rs756525122 Chr.3: 184852486-184852486 on Build GRCh38
rs774732839 Chr.3: 184852534-184852534 on Build GRCh38
rs774681527 Chr.3: 184852475-184852475 on Build GRCh38
rs896173873 Chr.3: 184852431-184852431 on Build GRCh38
rs778587238 Chr.3: 184852505-184852505 on Build GRCh38
rs188984274 Chr.3: 184852426-184852426 on Build GRCh38
rs761079092 Chr.3: 184852539-184852539 on Build GRCh38
rs766772396 Chr.3: 184852469-184852469 on Build GRCh38
rs866031348 Chr.3: 184852513-184852513 on Build GRCh38
rs577317442 Chr.3: 184852478-184852478 on Build GRCh38
rs553633908 Chr.3: 184852543-184852543 on Build GRCh38
rs370026889 Chr.3: 184852518-184852518 on Build GRCh38
rs749417221 Chr.3: 184852532-184852532 on Build GRCh38
rs200088304 Chr.3: 184852507-184852507 on Build GRCh38
rs763109717 Chr.3: 184852458-184852458 on Build GRCh38
rs753256589 Chr.3: 184852483-184852483 on Build GRCh38
rs376129060 Chr.3: 184852538-184852538 on Build GRCh38
rs781726133 Chr.3: 184852546-184852546 on Build GRCh38
rs558672776 Chr.3: 184852442-184852442 on Build GRCh38
rs753415075 Chr.3: 184852499-184852499 on Build GRCh38
rs756731589 Chr.3: 184852503-184852503 on Build GRCh38
rs768532730 Chr.3: 184852519-184852519 on Build GRCh38
rs141169650 Chr.3: 184852514-184852514 on Build GRCh38
rs373202010 Chr.3: 184852533-184852533 on Build GRCh38
rs773367236 Chr.3: 184852452-184852452 on Build GRCh38
rs764574647 Chr.3: 184852498-184852498 on Build GRCh38
rs764627867 Chr.3: 184852557-184852557 on Build GRCh38
rs776702852 Chr.3: 184852522-184852522 on Build GRCh38
rs761336402 Chr.3: 184852588-184852588 on Build GRCh38

View Full Product Details