Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150081803-150082062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS45
Entrez Gene ID
11311
Gene Name
vacuolar protein sorting 45 homolog
Gene Aliases
H1, H1VPS45, SCN5, VPS45A, VPS45B, VPS54A, VSP45, VSP45A
UniGene
Hs.443750
Gene Chromosome Location
Chr.1: 150067293-150145327 on Build GRCh38
Transcripts
NM_001279353.1 NM_001279354.1 NM_001279355.1 NM_007259.4 NR_103998.1 XM_017000164.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3801725 c.921A>G p.S307S Chr.1: 150081982-150081982 on Build GRCh38
895456 c.929A>G p.D310G Chr.1: 150081990-150081990 on Build GRCh38
5827494 c.565T>G p.F189V Chr.1: 150081941-150081941 on Build GRCh38
1333773 c.880T>G p.F294V Chr.1: 150081941-150081941 on Build GRCh38
3704944 c.532A>G p.I178V Chr.1: 150081908-150081908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782538917 Chr.1: 150081993-150081993 on Build GRCh38
rs782326494 Chr.1: 150081892-150081892 on Build GRCh38
rs782789330 Chr.1: 150081933-150081933 on Build GRCh38
rs1019641850 Chr.1: 150081876-150081876 on Build GRCh38
rs782583704 Chr.1: 150081987-150081987 on Build GRCh38
rs782654969 Chr.1: 150081994-150081994 on Build GRCh38
rs782284645 Chr.1: 150081841-150081841 on Build GRCh38
rs781988321 Chr.1: 150081852-150081852 on Build GRCh38
rs34757530 Chr.1: 150081878-150081878 on Build GRCh38
rs370189492 Chr.1: 150082008-150082008 on Build GRCh38
rs782467858 Chr.1: 150082018-150082018 on Build GRCh38
rs782079739 Chr.1: 150081853-150081853 on Build GRCh38
rs145286605 Chr.1: 150081941-150081941 on Build GRCh38
rs782476564 Chr.1: 150081849-150081849 on Build GRCh38
rs782739886 Chr.1: 150081959-150081959 on Build GRCh38
rs145369624 Chr.1: 150081991-150081991 on Build GRCh38
rs782455200 Chr.1: 150081979-150081979 on Build GRCh38
rs781916419 Chr.1: 150081907-150081907 on Build GRCh38
rs781867731 Chr.1: 150081956-150081956 on Build GRCh38
rs782391099 Chr.1: 150081843-150081843 on Build GRCh38
rs781829979 Chr.1: 150081967-150081967 on Build GRCh38
rs782164464 Chr.1: 150081918-150081918 on Build GRCh38
rs184193549 Chr.1: 150081958-150081958 on Build GRCh38

View Full Product Details