Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35675843-35676068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UPK1A
Entrez Gene ID
11045
Gene Name
uroplakin 1A
Gene Aliases
TSPAN21, UP1A, UPIA, UPKA
UniGene
Hs.159309
Gene Chromosome Location
Chr.19: 35666127-35678485 on Build GRCh38
Transcripts
NM_001281443.1 NM_007000.3 XM_017026225.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
165415 c.515C>T p.T172M Chr.19: 35675886-35675886 on Build GRCh38
5726385 c.513C>T p.F171F Chr.19: 35675884-35675884 on Build GRCh38
3532637 c.591C>T p.F197F Chr.19: 35675962-35675962 on Build GRCh38
3707061 c.526C>T p.R176W Chr.19: 35675897-35675897 on Build GRCh38
4553019 c.578G>A p.R193Q Chr.19: 35675949-35675949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764691019 Chr.19: 35675968-35675968 on Build GRCh38
rs768256456 Chr.19: 35675914-35675927 on Build GRCh38
rs563419319 Chr.19: 35675865-35675865 on Build GRCh38
rs371139752 Chr.19: 35675932-35675932 on Build GRCh38
rs374260140 Chr.19: 35675936-35675936 on Build GRCh38
rs767759607 Chr.19: 35675974-35675974 on Build GRCh38
rs771707160 Chr.19: 35675948-35675948 on Build GRCh38
rs757651035 Chr.19: 35675972-35675972 on Build GRCh38
rs770021966 Chr.19: 35676027-35676027 on Build GRCh38
rs542695953 Chr.19: 35675887-35675888 on Build GRCh38
rs746683190 Chr.19: 35675870-35675870 on Build GRCh38
rs751939866 Chr.19: 35675969-35675969 on Build GRCh38
rs563275041 Chr.19: 35676028-35676028 on Build GRCh38
rs746141008 Chr.19: 35676011-35676011 on Build GRCh38
rs757518845 Chr.19: 35675919-35675919 on Build GRCh38
rs766655398 Chr.19: 35675909-35675909 on Build GRCh38
rs555188398 Chr.19: 35675945-35675945 on Build GRCh38
rs751973417 Chr.19: 35675918-35675918 on Build GRCh38
rs755795192 Chr.19: 35675981-35675981 on Build GRCh38
rs745458744 Chr.19: 35675925-35675925 on Build GRCh38
rs146205685 Chr.19: 35675989-35675989 on Build GRCh38
rs780657748 Chr.19: 35675886-35675886 on Build GRCh38
rs770591821 Chr.19: 35675874-35675874 on Build GRCh38
rs200321267 Chr.19: 35675887-35675887 on Build GRCh38
rs866512502 Chr.19: 35675892-35675892 on Build GRCh38
rs761468255 Chr.19: 35675905-35675905 on Build GRCh38
rs377645091 Chr.19: 35675952-35675952 on Build GRCh38
rs764019699 Chr.19: 35676030-35676030 on Build GRCh38
rs772963912 Chr.19: 35676003-35676003 on Build GRCh38
rs775528882 Chr.19: 35675955-35675955 on Build GRCh38
rs768637606 Chr.19: 35675901-35675901 on Build GRCh38
rs896634037 Chr.19: 35675930-35675930 on Build GRCh38
rs753942126 Chr.19: 35675910-35675910 on Build GRCh38
rs181155606 Chr.19: 35676038-35676038 on Build GRCh38
rs762969376 Chr.19: 35675960-35675960 on Build GRCh38
rs779704385 Chr.19: 35675933-35675933 on Build GRCh38
rs141672780 Chr.19: 35675975-35675975 on Build GRCh38
rs954816562 Chr.19: 35675999-35675999 on Build GRCh38
rs780897710 Chr.19: 35675923-35675923 on Build GRCh38
rs748025807 Chr.19: 35675995-35675995 on Build GRCh38
rs771971555 Chr.19: 35676000-35676000 on Build GRCh38
rs148350758 Chr.19: 35675915-35675915 on Build GRCh38
rs377763111 Chr.19: 35675867-35675867 on Build GRCh38
rs961694135 Chr.19: 35675938-35675952 on Build GRCh38
rs201219777 Chr.19: 35675913-35675913 on Build GRCh38
rs774174868 Chr.19: 35675902-35675902 on Build GRCh38
rs752824114 Chr.19: 35676039-35676050 on Build GRCh38
rs35399814 Chr.19: 35675911-35675911 on Build GRCh38
rs941781822 Chr.19: 35676037-35676037 on Build GRCh38
rs888248707 Chr.19: 35675984-35675984 on Build GRCh38
rs778925384 Chr.19: 35675994-35675994 on Build GRCh38
rs779283933 Chr.19: 35676015-35676015 on Build GRCh38
rs749277609 Chr.19: 35675934-35675934 on Build GRCh38
rs535716709 Chr.19: 35675916-35675916 on Build GRCh38
rs1046058623 Chr.19: 35675920-35675920 on Build GRCh38
rs62109999 Chr.19: 35675950-35675950 on Build GRCh38
rs753709696 Chr.19: 35675890-35675891 on Build GRCh38
rs774979106 Chr.19: 35676034-35676034 on Build GRCh38
rs542534233 Chr.19: 35676042-35676042 on Build GRCh38
rs775581454 Chr.19: 35675897-35675897 on Build GRCh38
rs774118061 Chr.19: 35675943-35675943 on Build GRCh38
rs577633517 Chr.19: 35675988-35675988 on Build GRCh38
rs368710016 Chr.19: 35675898-35675898 on Build GRCh38
rs983286240 Chr.19: 35676009-35676009 on Build GRCh38
rs769359108 Chr.19: 35675888-35675888 on Build GRCh38
rs779804636 Chr.19: 35675987-35675987 on Build GRCh38
rs202063955 Chr.19: 35675949-35675949 on Build GRCh38
rs765344049 Chr.19: 35675953-35675953 on Build GRCh38

View Full Product Details