Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 110451736-110452005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGAG1
Entrez Gene ID
57529
Gene Name
retrotransposon gag domain containing 1
Gene Aliases
MAR9, MART9
UniGene
Hs.201071
Gene Chromosome Location
Chr.X: 110358101-110456334 on Build GRCh38
Transcripts
NM_020769.2 XM_011530998.2 XM_011530999.2 XM_017029694.1 XM_017029695.1

SNP Detail

SNP ID Chromosome Location
rs745650063 Chr.X: 110451786-110451786 on Build GRCh38
rs148882130 Chr.X: 110451825-110451825 on Build GRCh38
rs775538543 Chr.X: 110451798-110451798 on Build GRCh38
rs777204711 Chr.X: 110451916-110451916 on Build GRCh38
rs2073786 Chr.X: 110451826-110451826 on Build GRCh38
rs768005224 Chr.X: 110451806-110451806 on Build GRCh38
rs761304548 Chr.X: 110451811-110451811 on Build GRCh38
rs752773456 Chr.X: 110451948-110451948 on Build GRCh38
rs771759848 Chr.X: 110451904-110451904 on Build GRCh38
rs764442457 Chr.X: 110451954-110451954 on Build GRCh38
rs765764483 Chr.X: 110451941-110451941 on Build GRCh38
rs1025920250 Chr.X: 110451834-110451834 on Build GRCh38
rs191731262 Chr.X: 110451845-110451845 on Build GRCh38
rs772077898 Chr.X: 110451950-110451951 on Build GRCh38
rs780164731 Chr.X: 110451876-110451876 on Build GRCh38
rs766678813 Chr.X: 110451911-110451911 on Build GRCh38
rs754254608 Chr.X: 110451842-110451842 on Build GRCh38
rs1005978898 Chr.X: 110451764-110451764 on Build GRCh38
rs773798942 Chr.X: 110451889-110451889 on Build GRCh38
rs774068370 Chr.X: 110451809-110451809 on Build GRCh38
rs752820869 Chr.X: 110451857-110451857 on Build GRCh38
rs769906122 Chr.X: 110451793-110451793 on Build GRCh38
rs769672389 Chr.X: 110451872-110451872 on Build GRCh38
rs201596621 Chr.X: 110451933-110451933 on Build GRCh38
rs772817370 Chr.X: 110451910-110451910 on Build GRCh38
rs143680258 Chr.X: 110451951-110451951 on Build GRCh38
rs138209810 Chr.X: 110451858-110451858 on Build GRCh38
rs755104919 Chr.X: 110451844-110451844 on Build GRCh38
rs188623114 Chr.X: 110451780-110451780 on Build GRCh38
rs755987922 Chr.X: 110451965-110451965 on Build GRCh38
rs745699098 Chr.X: 110451871-110451871 on Build GRCh38
rs145500444 Chr.X: 110451805-110451805 on Build GRCh38
rs4314820 Chr.X: 110451838-110451838 on Build GRCh38
rs374873915 Chr.X: 110451781-110451781 on Build GRCh38
rs779648205 Chr.X: 110451966-110451966 on Build GRCh38
rs751782475 Chr.X: 110451960-110451960 on Build GRCh38
rs747681283 Chr.X: 110451895-110451895 on Build GRCh38
rs764520099 Chr.X: 110451859-110451859 on Build GRCh38
rs866842838 Chr.X: 110451936-110451936 on Build GRCh38
rs113549647 Chr.X: 110451800-110451800 on Build GRCh38
rs749124276 Chr.X: 110451888-110451888 on Build GRCh38
rs769776639 Chr.X: 110451820-110451820 on Build GRCh38
rs994816196 Chr.X: 110451821-110451821 on Build GRCh38

View Full Product Details