Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179955549-179955806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF130
Entrez Gene ID
55819
Gene Name
ring finger protein 130
Gene Aliases
G1RZFP, GOLIATH, GP
UniGene
Hs.484363
Gene Chromosome Location
Chr.5: 179911651-180072130 on Build GRCh38
Transcripts
NM_001280801.1 NM_018434.5 XM_011534593.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
482640 c.1257T>C p.F419F Chr.5: 179955657-179955657 on Build GRCh38
5469042 c.1245-4A>T p.? Chr.5: 179955673-179955673 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs566792978 Chr.5: 179955724-179955724 on Build GRCh38
rs761903856 Chr.5: 179955600-179955600 on Build GRCh38
rs751757696 Chr.5: 179955652-179955652 on Build GRCh38
rs773573028 Chr.5: 179955637-179955637 on Build GRCh38
rs775105339 Chr.5: 179955613-179955613 on Build GRCh38
rs976773577 Chr.5: 179955763-179955763 on Build GRCh38
rs367728987 Chr.5: 179955786-179955786 on Build GRCh38
rs905537562 Chr.5: 179955596-179955596 on Build GRCh38
rs748433375 Chr.5: 179955622-179955623 on Build GRCh38
rs367704518 Chr.5: 179955693-179955693 on Build GRCh38
rs746666668 Chr.5: 179955714-179955714 on Build GRCh38
rs762282750 Chr.5: 179955621-179955621 on Build GRCh38
rs1055856029 Chr.5: 179955716-179955716 on Build GRCh38
rs12332469 Chr.5: 179955594-179955594 on Build GRCh38
rs771535134 Chr.5: 179955694-179955694 on Build GRCh38
rs985737494 Chr.5: 179955737-179955737 on Build GRCh38
rs753600815 Chr.5: 179955684-179955684 on Build GRCh38
rs1011533311 Chr.5: 179955579-179955579 on Build GRCh38
rs767901987 Chr.5: 179955653-179955653 on Build GRCh38
rs752894077 Chr.5: 179955671-179955671 on Build GRCh38
rs778806016 Chr.5: 179955691-179955691 on Build GRCh38
rs773519031 Chr.5: 179955643-179955643 on Build GRCh38
rs894219646 Chr.5: 179955784-179955784 on Build GRCh38
rs554358717 Chr.5: 179955642-179955644 on Build GRCh38
rs14755 Chr.5: 179955597-179955597 on Build GRCh38
rs965367899 Chr.5: 179955752-179955752 on Build GRCh38
rs779581441 Chr.5: 179955707-179955707 on Build GRCh38
rs770041480 Chr.5: 179955647-179955647 on Build GRCh38
rs761036559 Chr.5: 179955695-179955695 on Build GRCh38
rs766924548 Chr.5: 179955651-179955651 on Build GRCh38
rs777441983 Chr.5: 179955682-179955682 on Build GRCh38
rs534138044 Chr.5: 179955787-179955787 on Build GRCh38
rs1018191887 Chr.5: 179955744-179955744 on Build GRCh38
rs765547987 Chr.5: 179955636-179955636 on Build GRCh38
rs756310208 Chr.5: 179955674-179955674 on Build GRCh38

View Full Product Details