Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44506774-44507003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERINC3
Entrez Gene ID
10955
Gene Name
serine incorporator 3
Gene Aliases
AIGP1, DIFF33, SBBI99, TDE, TDE1, TMS-1
UniGene
Hs.272168
Gene Chromosome Location
Chr.20: 44496221-44522116 on Build GRCh38
Transcripts
NM_006811.3 NM_198941.2 XM_017027601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7265450 c.634G>A p.A212T Chr.20: 44506976-44506976 on Build GRCh38
724497 c.634G>T p.A212S Chr.20: 44506976-44506976 on Build GRCh38
6775427 c.702C>T p.C234C Chr.20: 44506908-44506908 on Build GRCh38
3292917 c.674A>G p.Y225C Chr.20: 44506936-44506936 on Build GRCh38
6775424 c.732C>T p.N244N Chr.20: 44506878-44506878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771800103 Chr.20: 44506845-44506845 on Build GRCh38
rs761763639 Chr.20: 44506933-44506933 on Build GRCh38
rs778302835 Chr.20: 44506899-44506899 on Build GRCh38
rs150441257 Chr.20: 44506938-44506938 on Build GRCh38
rs375434063 Chr.20: 44506840-44506840 on Build GRCh38
rs202110058 Chr.20: 44506897-44506897 on Build GRCh38
rs761712355 Chr.20: 44506857-44506857 on Build GRCh38
rs563282440 Chr.20: 44506947-44506947 on Build GRCh38
rs775967540 Chr.20: 44506932-44506932 on Build GRCh38
rs781109565 Chr.20: 44506874-44506874 on Build GRCh38
rs752773259 Chr.20: 44506970-44506970 on Build GRCh38
rs199540220 Chr.20: 44506865-44506865 on Build GRCh38
rs200749563 Chr.20: 44506953-44506953 on Build GRCh38
rs138396148 Chr.20: 44506955-44506955 on Build GRCh38
rs148307225 Chr.20: 44506853-44506853 on Build GRCh38
rs750172297 Chr.20: 44506864-44506864 on Build GRCh38
rs768256372 Chr.20: 44506924-44506924 on Build GRCh38
rs760227907 Chr.20: 44506921-44506921 on Build GRCh38
rs374769277 Chr.20: 44506928-44506928 on Build GRCh38
rs902989165 Chr.20: 44506967-44506967 on Build GRCh38
rs765050040 Chr.20: 44506858-44506858 on Build GRCh38
rs777261763 Chr.20: 44506818-44506818 on Build GRCh38
rs755123999 Chr.20: 44506962-44506962 on Build GRCh38
rs200115653 Chr.20: 44506977-44506977 on Build GRCh38
rs750225048 Chr.20: 44506940-44506940 on Build GRCh38
rs781363126 Chr.20: 44506965-44506965 on Build GRCh38
rs748590093 Chr.20: 44506819-44506819 on Build GRCh38
rs796829781 Chr.20: 44506946-44506946 on Build GRCh38
rs202170652 Chr.20: 44506855-44506855 on Build GRCh38
rs750113502 Chr.20: 44506808-44506808 on Build GRCh38
rs760161466 Chr.20: 44506850-44506850 on Build GRCh38
rs762804409 Chr.20: 44506944-44506944 on Build GRCh38
rs1054838068 Chr.20: 44506826-44506826 on Build GRCh38
rs570011073 Chr.20: 44506832-44506832 on Build GRCh38
rs755145395 Chr.20: 44506872-44506872 on Build GRCh38
rs768689936 Chr.20: 44506951-44506951 on Build GRCh38
rs748729917 Chr.20: 44506896-44506896 on Build GRCh38
rs774873506 Chr.20: 44506846-44506846 on Build GRCh38
rs140439942 Chr.20: 44506915-44506915 on Build GRCh38
rs965532362 Chr.20: 44506936-44506936 on Build GRCh38
rs201595758 Chr.20: 44506912-44506912 on Build GRCh38
rs772584364 Chr.20: 44506805-44506807 on Build GRCh38
rs751726597 Chr.20: 44506866-44506866 on Build GRCh38
rs759290984 Chr.20: 44506844-44506844 on Build GRCh38
rs749603490 Chr.20: 44506907-44506907 on Build GRCh38
rs765104847 Chr.20: 44506939-44506939 on Build GRCh38
rs770330546 Chr.20: 44506821-44506821 on Build GRCh38

View Full Product Details