Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67184674-67184921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD3
Entrez Gene ID
4088
Gene Name
SMAD family member 3
Gene Aliases
HSPC193, HsT17436, JV15-2, LDS1C, LDS3, MADH3
UniGene
Hs.727986
Gene Chromosome Location
Chr.15: 67065698-67195195 on Build GRCh38
Transcripts
NM_001145102.1 NM_001145103.1 NM_001145104.1 NM_005902.3 XM_011521559.2 XM_011521560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5574980 c.950C>G p.S317C Chr.15: 67184805-67184805 on Build GRCh38
1588073 c.836G>A p.R279H Chr.15: 67184823-67184823 on Build GRCh38
6981419 c.943G>T p.V315F Chr.15: 67184798-67184798 on Build GRCh38
6918872 c.846G>A p.W282* Chr.15: 67184833-67184833 on Build GRCh38
6364213 c.784G>T p.E262* Chr.15: 67184771-67184771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768529580 Chr.15: 67184715-67184715 on Build GRCh38
rs142620875 Chr.15: 67184745-67184745 on Build GRCh38
rs771384141 Chr.15: 67184788-67184788 on Build GRCh38
rs747862625 Chr.15: 67184733-67184733 on Build GRCh38
rs769683236 Chr.15: 67184734-67184734 on Build GRCh38
rs863223742 Chr.15: 67184726-67184726 on Build GRCh38
rs863223744 Chr.15: 67184735-67184735 on Build GRCh38
rs769475136 Chr.15: 67184861-67184861 on Build GRCh38
rs74421858 Chr.15: 67184708-67184708 on Build GRCh38
rs773943578 Chr.15: 67184752-67184752 on Build GRCh38
rs967072552 Chr.15: 67184822-67184822 on Build GRCh38
rs150994304 Chr.15: 67184839-67184839 on Build GRCh38
rs759087344 Chr.15: 67184876-67184876 on Build GRCh38
rs767186122 Chr.15: 67184898-67184898 on Build GRCh38
rs754409301 Chr.15: 67184780-67184780 on Build GRCh38
rs757548494 Chr.15: 67184786-67184786 on Build GRCh38
rs745330707 Chr.15: 67184702-67184702 on Build GRCh38
rs748178271 Chr.15: 67184857-67184857 on Build GRCh38
rs750756638 Chr.15: 67184789-67184789 on Build GRCh38
rs139616052 Chr.15: 67184740-67184740 on Build GRCh38
rs772242555 Chr.15: 67184871-67184871 on Build GRCh38
rs951444228 Chr.15: 67184895-67184895 on Build GRCh38
rs1000537379 Chr.15: 67184694-67184694 on Build GRCh38
rs909496243 Chr.15: 67184885-67184885 on Build GRCh38
rs772995932 Chr.15: 67184869-67184869 on Build GRCh38
rs919935485 Chr.15: 67184890-67184890 on Build GRCh38
rs764656928 Chr.15: 67184777-67184777 on Build GRCh38
rs774180178 Chr.15: 67184875-67184875 on Build GRCh38
rs863223768 Chr.15: 67184797-67184797 on Build GRCh38
rs776577220 Chr.15: 67184723-67184723 on Build GRCh38
rs536824265 Chr.15: 67184877-67184877 on Build GRCh38
rs772040097 Chr.15: 67184695-67184695 on Build GRCh38
rs772730200 Chr.15: 67184739-67184739 on Build GRCh38
rs758586312 Chr.15: 67184823-67184823 on Build GRCh38
rs34188464 Chr.15: 67184860-67184860 on Build GRCh38
rs749052145 Chr.15: 67184870-67184870 on Build GRCh38
rs747102986 Chr.15: 67184712-67184712 on Build GRCh38
rs863223745 Chr.15: 67184802-67184802 on Build GRCh38
rs776999125 Chr.15: 67184901-67184901 on Build GRCh38
rs759140708 Chr.15: 67184767-67184767 on Build GRCh38
rs774470515 Chr.15: 67184845-67184845 on Build GRCh38
rs780274581 Chr.15: 67184836-67184836 on Build GRCh38
rs781081744 Chr.15: 67184854-67184854 on Build GRCh38
rs863223754 Chr.15: 67184845-67184846 on Build GRCh38

View Full Product Details