Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 53042924-53043177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLFM4
Entrez Gene ID
10562
Gene Name
olfactomedin 4
Gene Aliases
GC1, GW112, OLM4, OlfD, UNQ362, bA209J19.1, hGC-1, hOLfD
UniGene
Hs.508113
Gene Chromosome Location
Chr.13: 53028741-53052061 on Build GRCh38
Transcripts
NM_006418.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
948128 c.632C>T p.A211V Chr.13: 53043166-53043166 on Build GRCh38
1367496 c.643G>T p.E215* Chr.13: 53043177-53043177 on Build GRCh38
3469369 c.586C>T p.L196F Chr.13: 53043120-53043120 on Build GRCh38
4912390 c.600G>A p.K200K Chr.13: 53043134-53043134 on Build GRCh38
948127 c.586C>A p.L196I Chr.13: 53043120-53043120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750934849 Chr.13: 53043147-53043147 on Build GRCh38
rs764161531 Chr.13: 53043101-53043101 on Build GRCh38
rs371258053 Chr.13: 53043124-53043124 on Build GRCh38
rs751838760 Chr.13: 53043115-53043115 on Build GRCh38
rs760253975 Chr.13: 53043067-53043067 on Build GRCh38
rs776287962 Chr.13: 53043109-53043109 on Build GRCh38
rs768010019 Chr.13: 53043125-53043125 on Build GRCh38
rs879246259 Chr.13: 53043102-53043102 on Build GRCh38
rs199556573 Chr.13: 53043112-53043112 on Build GRCh38
rs960296771 Chr.13: 53043045-53043045 on Build GRCh38
rs759133822 Chr.13: 53043110-53043110 on Build GRCh38
rs1036360529 Chr.13: 53042980-53042980 on Build GRCh38
rs879045699 Chr.13: 53043100-53043100 on Build GRCh38
rs773716458 Chr.13: 53043069-53043069 on Build GRCh38
rs878920006 Chr.13: 53043101-53043102 on Build GRCh38
rs910034394 Chr.13: 53043081-53043081 on Build GRCh38
rs202159204 Chr.13: 53043051-53043051 on Build GRCh38
rs200611659 Chr.13: 53043055-53043055 on Build GRCh38
rs1026277496 Chr.13: 53043033-53043033 on Build GRCh38
rs780024062 Chr.13: 53043160-53043160 on Build GRCh38
rs753801493 Chr.13: 53043145-53043145 on Build GRCh38
rs756131102 Chr.13: 53043156-53043156 on Build GRCh38
rs765988494 Chr.13: 53043090-53043090 on Build GRCh38

View Full Product Details