Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30086434-30086661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX6
Entrez Gene ID
6911
Gene Name
T-box 6
Gene Aliases
SCDO5
UniGene
Hs.198301
Gene Chromosome Location
Chr.16: 30085793-30091919 on Build GRCh38
Transcripts
NM_004608.3 XM_005255523.2 XM_011545926.2 XM_017023614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5510797 c.1003G>A p.A335T Chr.16: 30086606-30086606 on Build GRCh38
6279205 c.991G>A p.G331R Chr.16: 30086618-30086618 on Build GRCh38
7393230 c.982C>G p.P328A Chr.16: 30086627-30086627 on Build GRCh38
4333265 c.1073G>A p.G358E Chr.16: 30086536-30086536 on Build GRCh38
3888387 c.975A>G p.E325E Chr.16: 30086634-30086634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776075571 Chr.16: 30086458-30086458 on Build GRCh38
rs555486285 Chr.16: 30086519-30086519 on Build GRCh38
rs746046440 Chr.16: 30086561-30086561 on Build GRCh38
rs757310327 Chr.16: 30086624-30086624 on Build GRCh38
rs377659260 Chr.16: 30086589-30086589 on Build GRCh38
rs1013957062 Chr.16: 30086595-30086595 on Build GRCh38
rs778894075 Chr.16: 30086625-30086625 on Build GRCh38
rs756635841 Chr.16: 30086509-30086509 on Build GRCh38
rs199768549 Chr.16: 30086626-30086626 on Build GRCh38
rs901673570 Chr.16: 30086553-30086553 on Build GRCh38
rs764847381 Chr.16: 30086480-30086480 on Build GRCh38
rs771855960 Chr.16: 30086568-30086568 on Build GRCh38
rs768520466 Chr.16: 30086587-30086587 on Build GRCh38
rs780222043 Chr.16: 30086629-30086629 on Build GRCh38
rs764082046 Chr.16: 30086457-30086457 on Build GRCh38
rs567311252 Chr.16: 30086601-30086601 on Build GRCh38
rs545301471 Chr.16: 30086619-30086619 on Build GRCh38
rs553009645 Chr.16: 30086594-30086594 on Build GRCh38
rs761253380 Chr.16: 30086591-30086591 on Build GRCh38
rs775324500 Chr.16: 30086571-30086571 on Build GRCh38
rs1049176622 Chr.16: 30086498-30086498 on Build GRCh38
rs200023167 Chr.16: 30086548-30086548 on Build GRCh38
rs368712218 Chr.16: 30086598-30086598 on Build GRCh38
rs779589978 Chr.16: 30086499-30086500 on Build GRCh38
rs753179563 Chr.16: 30086607-30086607 on Build GRCh38
rs963515700 Chr.16: 30086622-30086622 on Build GRCh38
rs746833997 Chr.16: 30086635-30086635 on Build GRCh38
rs536875824 Chr.16: 30086497-30086497 on Build GRCh38
rs755426985 Chr.16: 30086482-30086482 on Build GRCh38
rs928797124 Chr.16: 30086476-30086476 on Build GRCh38
rs761069288 Chr.16: 30086610-30086610 on Build GRCh38
rs374335976 Chr.16: 30086615-30086615 on Build GRCh38
rs866620210 Chr.16: 30086569-30086569 on Build GRCh38
rs750047944 Chr.16: 30086481-30086481 on Build GRCh38
rs149105120 Chr.16: 30086599-30086599 on Build GRCh38
rs79360759 Chr.16: 30086547-30086547 on Build GRCh38
rs767928733 Chr.16: 30086488-30086488 on Build GRCh38
rs779126710 Chr.16: 30086558-30086558 on Build GRCh38
rs977584914 Chr.16: 30086453-30086453 on Build GRCh38
rs1029088576 Chr.16: 30086580-30086580 on Build GRCh38
rs746806888 Chr.16: 30086582-30086582 on Build GRCh38
rs376623017 Chr.16: 30086489-30086489 on Build GRCh38
rs947755293 Chr.16: 30086494-30086494 on Build GRCh38
rs766116040 Chr.16: 30086606-30086606 on Build GRCh38
rs943164118 Chr.16: 30086504-30086504 on Build GRCh38
rs761359356 Chr.16: 30086461-30086461 on Build GRCh38
rs758689411 Chr.16: 30086627-30086627 on Build GRCh38
rs762635226 Chr.16: 30086602-30086602 on Build GRCh38
rs764553748 Chr.16: 30086618-30086618 on Build GRCh38
rs746571204 Chr.16: 30086585-30086586 on Build GRCh38

View Full Product Details