Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65530565-65530836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HACD3
Entrez Gene ID
51495
Gene Name
3-hydroxyacyl-CoA dehydratase 3
Gene Aliases
B-IND1, BIND1, HSPC121, PTPLAD1
UniGene
Hs.512973
Gene Chromosome Location
Chr.15: 65530489-65578355 on Build GRCh38
Transcripts
NM_016395.2 XM_011521664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1222508 c.6G>T p.E2D Chr.15: 65530637-65530637 on Build GRCh38
4389469 c.1A>T p.M1L Chr.15: 65530632-65530632 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs527249644 Chr.15: 65530789-65530789 on Build GRCh38
rs372174820 Chr.15: 65530708-65530708 on Build GRCh38
rs750303074 Chr.15: 65530744-65530744 on Build GRCh38
rs1014170975 Chr.15: 65530696-65530696 on Build GRCh38
rs768389220 Chr.15: 65530594-65530594 on Build GRCh38
rs779678571 Chr.15: 65530757-65530757 on Build GRCh38
rs951720861 Chr.15: 65530802-65530802 on Build GRCh38
rs964807409 Chr.15: 65530797-65530797 on Build GRCh38
rs1045716059 Chr.15: 65530720-65530720 on Build GRCh38
rs995884676 Chr.15: 65530799-65530799 on Build GRCh38
rs764397884 Chr.15: 65530700-65530700 on Build GRCh38
rs746580673 Chr.15: 65530589-65530589 on Build GRCh38
rs1054853487 Chr.15: 65530716-65530716 on Build GRCh38
rs747874412 Chr.15: 65530767-65530767 on Build GRCh38
rs767934966 Chr.15: 65530615-65530615 on Build GRCh38
rs1043536952 Chr.15: 65530604-65530604 on Build GRCh38
rs374497608 Chr.15: 65530688-65530688 on Build GRCh38
rs1045798450 Chr.15: 65530617-65530617 on Build GRCh38
rs949856058 Chr.15: 65530608-65530608 on Build GRCh38
rs774990431 Chr.15: 65530606-65530606 on Build GRCh38
rs758340384 Chr.15: 65530752-65530752 on Build GRCh38
rs1027713733 Chr.15: 65530800-65530800 on Build GRCh38
rs754049796 Chr.15: 65530712-65530712 on Build GRCh38
rs1008360926 Chr.15: 65530772-65530772 on Build GRCh38
rs35333624 Chr.15: 65530668-65530669 on Build GRCh38
rs936197425 Chr.15: 65530631-65530631 on Build GRCh38
rs1018745267 Chr.15: 65530787-65530787 on Build GRCh38
rs372231534 Chr.15: 65530758-65530758 on Build GRCh38
rs1053345731 Chr.15: 65530680-65530680 on Build GRCh38
rs775957220 Chr.15: 65530595-65530595 on Build GRCh38
rs775926178 Chr.15: 65530682-65530682 on Build GRCh38
rs34736092 Chr.15: 65530730-65530730 on Build GRCh38
rs765358702 Chr.15: 65530733-65530733 on Build GRCh38
rs780862891 Chr.15: 65530765-65530765 on Build GRCh38
rs777968493 Chr.15: 65530765-65530768 on Build GRCh38
rs567976049 Chr.15: 65530762-65530762 on Build GRCh38
rs757326229 Chr.15: 65530725-65530725 on Build GRCh38
rs377113434 Chr.15: 65530611-65530611 on Build GRCh38
rs1020520852 Chr.15: 65530763-65530763 on Build GRCh38
rs761548750 Chr.15: 65530600-65530600 on Build GRCh38

View Full Product Details