Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83038407-83038664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE3D
Entrez Gene ID
90025
Gene Name
ubiquitin protein ligase E3D
Gene Aliases
C6orf157, H10BH, UBE2CBP, YJR141W
UniGene
Hs.148609
Gene Chromosome Location
Chr.6: 82880834-83065841 on Build GRCh38
Transcripts
NM_001304437.1 NM_198920.2 XM_011536238.2 XM_011536239.2 XM_011536240.1 XM_011536241.2 XM_011536242.1 XM_017011458.1 XM_017011459.1 XM_017011460.1 XM_017011461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6107659 c.605A>T p.K202M Chr.6: 83038478-83038478 on Build GRCh38
1081997 c.658G>A p.V220M Chr.6: 83038425-83038425 on Build GRCh38
743012 c.649G>A p.G217R Chr.6: 83038434-83038434 on Build GRCh38
743011 c.621A>G p.V207V Chr.6: 83038462-83038462 on Build GRCh38
6565172 c.598-6C>G p.? Chr.6: 83038491-83038491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755723328 Chr.6: 83038494-83038494 on Build GRCh38
rs1037958933 Chr.6: 83038572-83038572 on Build GRCh38
rs754110427 Chr.6: 83038447-83038447 on Build GRCh38
rs979364051 Chr.6: 83038575-83038575 on Build GRCh38
rs760811107 Chr.6: 83038443-83038443 on Build GRCh38
rs199505546 Chr.6: 83038489-83038489 on Build GRCh38
rs140596068 Chr.6: 83038437-83038437 on Build GRCh38
rs993755928 Chr.6: 83038621-83038621 on Build GRCh38
rs767927719 Chr.6: 83038433-83038433 on Build GRCh38
rs751315357 Chr.6: 83038453-83038453 on Build GRCh38
rs370988553 Chr.6: 83038431-83038431 on Build GRCh38
rs142837467 Chr.6: 83038451-83038451 on Build GRCh38
rs967668460 Chr.6: 83038547-83038547 on Build GRCh38
rs764128931 Chr.6: 83038445-83038445 on Build GRCh38
rs188165642 Chr.6: 83038520-83038520 on Build GRCh38
rs781332051 Chr.6: 83038482-83038482 on Build GRCh38
rs118061057 Chr.6: 83038632-83038632 on Build GRCh38
rs898043594 Chr.6: 83038601-83038601 on Build GRCh38
rs942150704 Chr.6: 83038530-83038530 on Build GRCh38
rs1023598780 Chr.6: 83038458-83038458 on Build GRCh38
rs754744091 Chr.6: 83038468-83038468 on Build GRCh38
rs906049092 Chr.6: 83038462-83038462 on Build GRCh38
rs1020622428 Chr.6: 83038518-83038518 on Build GRCh38
rs748793633 Chr.6: 83038522-83038522 on Build GRCh38
rs777392704 Chr.6: 83038496-83038496 on Build GRCh38
rs147405452 Chr.6: 83038452-83038452 on Build GRCh38

View Full Product Details