Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 118281831-118282092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS12
Entrez Gene ID
8492
Gene Name
protease, serine 12
Gene Aliases
BSSP-3, BSSP3, MRT1
UniGene
Hs.445857
Gene Chromosome Location
Chr.4: 118280038-118353042 on Build GRCh38
Transcripts
NM_003619.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5038819 c.2542_2543insG p.Y848fs*1 Chr.4: 118282021-118282022 on Build GRCh38
6341387 c.2502C>G p.P834P Chr.4: 118282062-118282062 on Build GRCh38
5079595 c.2545_2546insTATG p.G849fs*>29 Chr.4: 118282018-118282019 on Build GRCh38
5036975 c.2475C>T p.S825S Chr.4: 118282089-118282089 on Build GRCh38
420231 c.2564C>A p.S855Y Chr.4: 118282000-118282000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777770002 Chr.4: 118281930-118281930 on Build GRCh38
rs528081738 Chr.4: 118282070-118282070 on Build GRCh38
rs778245197 Chr.4: 118282015-118282015 on Build GRCh38
rs773463725 Chr.4: 118282021-118282022 on Build GRCh38
rs963868288 Chr.4: 118282024-118282024 on Build GRCh38
rs878982970 Chr.4: 118281922-118281922 on Build GRCh38
rs957912978 Chr.4: 118282001-118282001 on Build GRCh38
rs777025114 Chr.4: 118282018-118282018 on Build GRCh38
rs922467877 Chr.4: 118282056-118282056 on Build GRCh38
rs763434880 Chr.4: 118282023-118282023 on Build GRCh38
rs575750897 Chr.4: 118281951-118281951 on Build GRCh38
rs139833696 Chr.4: 118282061-118282061 on Build GRCh38
rs999133953 Chr.4: 118281989-118281989 on Build GRCh38
rs745366732 Chr.4: 118281891-118281898 on Build GRCh38
rs771746888 Chr.4: 118281916-118281924 on Build GRCh38
rs745984674 Chr.4: 118281984-118281984 on Build GRCh38
rs368847295 Chr.4: 118281898-118281898 on Build GRCh38
rs532143815 Chr.4: 118282047-118282047 on Build GRCh38
rs1014796300 Chr.4: 118281876-118281876 on Build GRCh38
rs766050375 Chr.4: 118282032-118282032 on Build GRCh38
rs754830727 Chr.4: 118281918-118281918 on Build GRCh38
rs368413746 Chr.4: 118282023-118282023 on Build GRCh38
rs777254934 Chr.4: 118281938-118281939 on Build GRCh38
rs774536447 Chr.4: 118281969-118281969 on Build GRCh38
rs556096183 Chr.4: 118281899-118281899 on Build GRCh38
rs903423887 Chr.4: 118281960-118281960 on Build GRCh38
rs74733027 Chr.4: 118282014-118282014 on Build GRCh38
rs145028190 Chr.4: 118281863-118281863 on Build GRCh38
rs765573893 Chr.4: 118281981-118281981 on Build GRCh38
rs765853490 Chr.4: 118281914-118281914 on Build GRCh38
rs749129340 Chr.4: 118282062-118282062 on Build GRCh38
rs200516485 Chr.4: 118282011-118282011 on Build GRCh38
rs762443507 Chr.4: 118281944-118281946 on Build GRCh38
rs372672235 Chr.4: 118282012-118282012 on Build GRCh38
rs747833223 Chr.4: 118281928-118281928 on Build GRCh38
rs745865957 Chr.4: 118282068-118282068 on Build GRCh38
rs751109607 Chr.4: 118281917-118281917 on Build GRCh38
rs936060513 Chr.4: 118282006-118282006 on Build GRCh38
rs145964630 Chr.4: 118282029-118282029 on Build GRCh38
rs886059033 Chr.4: 118281860-118281861 on Build GRCh38
rs17594503 Chr.4: 118282066-118282066 on Build GRCh38
rs1024515027 Chr.4: 118281903-118281903 on Build GRCh38
rs747524139 Chr.4: 118282072-118282072 on Build GRCh38
rs771322098 Chr.4: 118281968-118281968 on Build GRCh38
rs1033282830 Chr.4: 118281991-118281991 on Build GRCh38
rs558897637 Chr.4: 118282067-118282067 on Build GRCh38
rs754527511 Chr.4: 118282044-118282044 on Build GRCh38
rs138882049 Chr.4: 118281996-118281996 on Build GRCh38
rs781210570 Chr.4: 118281921-118281921 on Build GRCh38
rs577137531 Chr.4: 118282020-118282020 on Build GRCh38
rs756054852 Chr.4: 118282060-118282060 on Build GRCh38
rs751020682 Chr.4: 118282039-118282039 on Build GRCh38
rs767005194 Chr.4: 118282046-118282046 on Build GRCh38
rs989169715 Chr.4: 118282007-118282007 on Build GRCh38

View Full Product Details