Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140300491-140300744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PFDN1
Entrez Gene ID
5201
Gene Name
prefoldin subunit 1
Gene Aliases
PDF, PFD1
UniGene
Hs.483564
Gene Chromosome Location
Chr.5: 140245050-140303141 on Build GRCh38
Transcripts
NM_002622.4 XM_005268465.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1433533 c.119C>T p.T40M Chr.5: 140300497-140300497 on Build GRCh38
3768154 c.101T>G p.I34S Chr.5: 140300515-140300515 on Build GRCh38
1433534 c.88G>A p.A30T Chr.5: 140300528-140300528 on Build GRCh38
6881712 c.41C>T p.T14I Chr.5: 140300575-140300575 on Build GRCh38
3826890 c.99G>A p.Q33Q Chr.5: 140300517-140300517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749919941 Chr.5: 140300528-140300528 on Build GRCh38
rs779611397 Chr.5: 140300540-140300540 on Build GRCh38
rs753205604 Chr.5: 140300520-140300520 on Build GRCh38
rs748027033 Chr.5: 140300577-140300577 on Build GRCh38
rs943812220 Chr.5: 140300611-140300611 on Build GRCh38
rs907534377 Chr.5: 140300695-140300695 on Build GRCh38
rs918500446 Chr.5: 140300638-140300638 on Build GRCh38
rs377128367 Chr.5: 140300526-140300526 on Build GRCh38
rs746666692 Chr.5: 140300544-140300544 on Build GRCh38
rs767794566 Chr.5: 140300519-140300519 on Build GRCh38
rs769842902 Chr.5: 140300609-140300609 on Build GRCh38
rs749314513 Chr.5: 140300629-140300629 on Build GRCh38
rs754766534 Chr.5: 140300554-140300554 on Build GRCh38
rs1048518773 Chr.5: 140300671-140300671 on Build GRCh38
rs757966331 Chr.5: 140300529-140300529 on Build GRCh38
rs780821305 Chr.5: 140300555-140300555 on Build GRCh38
rs529125442 Chr.5: 140300686-140300686 on Build GRCh38
rs905559192 Chr.5: 140300674-140300674 on Build GRCh38
rs771149896 Chr.5: 140300632-140300632 on Build GRCh38
rs773351261 Chr.5: 140300616-140300616 on Build GRCh38
rs937018448 Chr.5: 140300675-140300675 on Build GRCh38
rs974234222 Chr.5: 140300620-140300620 on Build GRCh38
rs931285948 Chr.5: 140300654-140300654 on Build GRCh38
rs756545903 Chr.5: 140300522-140300522 on Build GRCh38
rs768225470 Chr.5: 140300587-140300588 on Build GRCh38

View Full Product Details