Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145141344-145141603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUD4
Entrez Gene ID
54726
Gene Name
OTU deubiquitinase 4
Gene Aliases
DUBA6, HIN1, HSHIN1
UniGene
Hs.270851
Gene Chromosome Location
Chr.4: 145133650-145180019 on Build GRCh38
Transcripts
NM_001102653.1 XM_005263079.4 XM_011532040.2 XM_011532041.2 XM_011532042.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
283657 c.1774A>G p.T592A Chr.4: 145141493-145141493 on Build GRCh38
1218999 c.1676C>T p.T559I Chr.4: 145141591-145141591 on Build GRCh38
5508683 c.1834C>A p.R612R Chr.4: 145141433-145141433 on Build GRCh38
3669213 c.1870A>G p.T624A Chr.4: 145141592-145141592 on Build GRCh38
7298541 c.2051C>T p.S684L Chr.4: 145141411-145141411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007010097 Chr.4: 145141552-145141552 on Build GRCh38
rs778577405 Chr.4: 145141485-145141485 on Build GRCh38
rs769425500 Chr.4: 145141538-145141538 on Build GRCh38
rs768082960 Chr.4: 145141524-145141524 on Build GRCh38
rs767589647 Chr.4: 145141458-145141458 on Build GRCh38
rs756301844 Chr.4: 145141480-145141480 on Build GRCh38
rs201690512 Chr.4: 145141564-145141564 on Build GRCh38
rs772065996 Chr.4: 145141389-145141389 on Build GRCh38
rs373613696 Chr.4: 145141398-145141398 on Build GRCh38
rs766942382 Chr.4: 145141448-145141448 on Build GRCh38
rs757029657 Chr.4: 145141373-145141373 on Build GRCh38
rs746914323 Chr.4: 145141518-145141518 on Build GRCh38
rs200084041 Chr.4: 145141422-145141422 on Build GRCh38
rs752881863 Chr.4: 145141472-145141472 on Build GRCh38
rs144732315 Chr.4: 145141553-145141553 on Build GRCh38
rs370095686 Chr.4: 145141580-145141580 on Build GRCh38
rs942104714 Chr.4: 145141414-145141414 on Build GRCh38
rs746577185 Chr.4: 145141410-145141410 on Build GRCh38
rs201743316 Chr.4: 145141571-145141571 on Build GRCh38
rs918045760 Chr.4: 145141383-145141383 on Build GRCh38
rs113829733 Chr.4: 145141575-145141575 on Build GRCh38
rs774868060 Chr.4: 145141555-145141555 on Build GRCh38
rs147236665 Chr.4: 145141451-145141451 on Build GRCh38
rs772850045 Chr.4: 145141543-145141543 on Build GRCh38
rs778717069 Chr.4: 145141386-145141386 on Build GRCh38
rs143751740 Chr.4: 145141420-145141420 on Build GRCh38
rs768192851 Chr.4: 145141413-145141413 on Build GRCh38
rs752755532 Chr.4: 145141581-145141581 on Build GRCh38
rs763369177 Chr.4: 145141551-145141551 on Build GRCh38
rs773572635 Chr.4: 145141433-145141433 on Build GRCh38
rs765546129 Chr.4: 145141428-145141428 on Build GRCh38
rs755088734 Chr.4: 145141453-145141453 on Build GRCh38
rs75726118 Chr.4: 145141530-145141530 on Build GRCh38
rs140724824 Chr.4: 145141516-145141516 on Build GRCh38
rs377468177 Chr.4: 145141395-145141395 on Build GRCh38
rs758252968 Chr.4: 145141505-145141505 on Build GRCh38
rs927416676 Chr.4: 145141486-145141486 on Build GRCh38
rs372757053 Chr.4: 145141490-145141490 on Build GRCh38
rs763458478 Chr.4: 145141443-145141443 on Build GRCh38

View Full Product Details