Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44570454-44570705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPG11
Entrez Gene ID
80208
Gene Name
spastic paraplegia 11 (autosomal recessive)
Gene Aliases
ALS5, CMT2X, KIAA1840
UniGene
Hs.656271
Gene Chromosome Location
Chr.15: 44562696-44663678 on Build GRCh38
Transcripts
NM_001160227.1 NM_025137.3 XM_006720700.1 XM_017022634.1 XM_017022636.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6348381 c.6426C>A p.A2142A Chr.15: 44570576-44570576 on Build GRCh38
6250698 c.6381C>A p.T2127T Chr.15: 44570621-44570621 on Build GRCh38
3769594 c.6387G>A p.T2129T Chr.15: 44570615-44570615 on Build GRCh38
5516694 c.6386C>T p.T2129M Chr.15: 44570616-44570616 on Build GRCh38
3956654 c.6468T>C p.Y2156Y Chr.15: 44570534-44570534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770736243 Chr.15: 44570507-44570507 on Build GRCh38
rs376146440 Chr.15: 44570491-44570491 on Build GRCh38
rs543344637 Chr.15: 44570544-44570544 on Build GRCh38
rs759378435 Chr.15: 44570527-44570527 on Build GRCh38
rs1015008074 Chr.15: 44570681-44570681 on Build GRCh38
rs312262780 Chr.15: 44570521-44570521 on Build GRCh38
rs375608115 Chr.15: 44570616-44570616 on Build GRCh38
rs574960359 Chr.15: 44570512-44570512 on Build GRCh38
rs769898852 Chr.15: 44570593-44570593 on Build GRCh38
rs748324376 Chr.15: 44570517-44570519 on Build GRCh38
rs755071134 Chr.15: 44570503-44570504 on Build GRCh38
rs763676031 Chr.15: 44570550-44570550 on Build GRCh38
rs773909173 Chr.15: 44570622-44570622 on Build GRCh38
rs751684367 Chr.15: 44570493-44570493 on Build GRCh38
rs768834747 Chr.15: 44570488-44570488 on Build GRCh38
rs775209503 Chr.15: 44570611-44570611 on Build GRCh38
rs747227001 Chr.15: 44570481-44570481 on Build GRCh38
rs150121550 Chr.15: 44570615-44570615 on Build GRCh38
rs770158083 Chr.15: 44570641-44570641 on Build GRCh38
rs141035224 Chr.15: 44570578-44570578 on Build GRCh38
rs763355260 Chr.15: 44570678-44570682 on Build GRCh38
rs774157270 Chr.15: 44570513-44570513 on Build GRCh38
rs750928941 Chr.15: 44570565-44570565 on Build GRCh38
rs764834822 Chr.15: 44570621-44570621 on Build GRCh38
rs780549081 Chr.15: 44570570-44570570 on Build GRCh38
rs758266744 Chr.15: 44570682-44570682 on Build GRCh38
rs758895071 Chr.15: 44570567-44570567 on Build GRCh38
rs559725145 Chr.15: 44570475-44570475 on Build GRCh38
rs779857193 Chr.15: 44570685-44570685 on Build GRCh38
rs200283964 Chr.15: 44570592-44570592 on Build GRCh38
rs761269744 Chr.15: 44570618-44570618 on Build GRCh38
rs745820206 Chr.15: 44570675-44570675 on Build GRCh38
rs565057074 Chr.15: 44570671-44570671 on Build GRCh38
rs370189964 Chr.15: 44570596-44570596 on Build GRCh38
rs373897153 Chr.15: 44570555-44570555 on Build GRCh38
rs369450223 Chr.15: 44570674-44570674 on Build GRCh38
rs146240471 Chr.15: 44570534-44570534 on Build GRCh38
rs368605117 Chr.15: 44570642-44570642 on Build GRCh38
rs139650852 Chr.15: 44570542-44570542 on Build GRCh38
rs312262779 Chr.15: 44570551-44570551 on Build GRCh38
rs751841565 Chr.15: 44570572-44570572 on Build GRCh38
rs952927034 Chr.15: 44570579-44570579 on Build GRCh38
rs771913148 Chr.15: 44570607-44570607 on Build GRCh38
rs762403160 Chr.15: 44570551-44570551 on Build GRCh38
rs923174572 Chr.15: 44570668-44570668 on Build GRCh38
rs766851227 Chr.15: 44570650-44570650 on Build GRCh38
rs376753565 Chr.15: 44570506-44570506 on Build GRCh38
rs781072777 Chr.15: 44570489-44570489 on Build GRCh38
rs751929116 Chr.15: 44570655-44570655 on Build GRCh38
rs767684146 Chr.15: 44570666-44570666 on Build GRCh38
rs745801884 Chr.15: 44570600-44570600 on Build GRCh38
rs752935771 Chr.15: 44570667-44570667 on Build GRCh38
rs982768284 Chr.15: 44570589-44570589 on Build GRCh38
rs777817631 Chr.15: 44570673-44570673 on Build GRCh38
rs755253734 Chr.15: 44570664-44570664 on Build GRCh38
rs771827244 Chr.15: 44570541-44570541 on Build GRCh38

View Full Product Details