Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3828013-3828238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOG
Entrez Gene ID
391
Gene Name
ras homolog family member G
Gene Aliases
ARHG
UniGene
Hs.501728
Gene Chromosome Location
Chr.11: 3826978-3840983 on Build GRCh38
Transcripts
NM_001665.3 XM_005252916.2 XM_017017719.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4721583 c.83T>C p.F28S Chr.11: 3828056-3828056 on Build GRCh38
1717608 c.34G>A p.G12R Chr.11: 3828105-3828105 on Build GRCh38
1746225 c.21G>T p.V7V Chr.11: 3828118-3828118 on Build GRCh38
1604529 c.125C>T p.A42V Chr.11: 3828014-3828014 on Build GRCh38
926827 c.79G>A p.A27T Chr.11: 3828060-3828060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766457887 Chr.11: 3828109-3828109 on Build GRCh38
rs748092390 Chr.11: 3828146-3828146 on Build GRCh38
rs745411648 Chr.11: 3828186-3828186 on Build GRCh38
rs141646821 Chr.11: 3828121-3828121 on Build GRCh38
rs748728475 Chr.11: 3828176-3828177 on Build GRCh38
rs901323177 Chr.11: 3828158-3828158 on Build GRCh38
rs770661565 Chr.11: 3828171-3828171 on Build GRCh38
rs764657068 Chr.11: 3828043-3828043 on Build GRCh38
rs776225932 Chr.11: 3828037-3828037 on Build GRCh38
rs924524842 Chr.11: 3828094-3828094 on Build GRCh38
rs985680774 Chr.11: 3828169-3828169 on Build GRCh38
rs768185722 Chr.11: 3828179-3828182 on Build GRCh38
rs770573794 Chr.11: 3828034-3828034 on Build GRCh38
rs750677069 Chr.11: 3828065-3828065 on Build GRCh38
rs962877052 Chr.11: 3828180-3828180 on Build GRCh38
rs778753436 Chr.11: 3828139-3828139 on Build GRCh38
rs1025814558 Chr.11: 3828191-3828191 on Build GRCh38
rs1018137274 Chr.11: 3828176-3828176 on Build GRCh38
rs372321496 Chr.11: 3828181-3828181 on Build GRCh38
rs977311811 Chr.11: 3828082-3828082 on Build GRCh38
rs763579274 Chr.11: 3828040-3828040 on Build GRCh38
rs767911108 Chr.11: 3828061-3828061 on Build GRCh38
rs756262131 Chr.11: 3828103-3828103 on Build GRCh38
rs746656564 Chr.11: 3828168-3828168 on Build GRCh38
rs1054186579 Chr.11: 3828115-3828115 on Build GRCh38
rs762134439 Chr.11: 3828060-3828060 on Build GRCh38
rs949877516 Chr.11: 3828076-3828076 on Build GRCh38
rs918678475 Chr.11: 3828195-3828195 on Build GRCh38
rs893844189 Chr.11: 3828120-3828120 on Build GRCh38
rs754937488 Chr.11: 3828124-3828124 on Build GRCh38
rs189821024 Chr.11: 3828214-3828214 on Build GRCh38
rs777561222 Chr.11: 3828148-3828148 on Build GRCh38
rs758182032 Chr.11: 3828147-3828147 on Build GRCh38
rs774893077 Chr.11: 3828046-3828046 on Build GRCh38

View Full Product Details