Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58571884-58572157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX14
Entrez Gene ID
56155
Gene Name
testis expressed 14, intercellular bridge forming factor
Gene Aliases
CT113
UniGene
Hs.390221
Gene Chromosome Location
Chr.17: 58556677-58692055 on Build GRCh38
Transcripts
NM_001201457.1 NM_031272.4 NM_198393.3 XM_011525029.2 XM_017024860.1 XM_017024861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6988287 c.3549C>A p.C1183* Chr.17: 58572089-58572089 on Build GRCh38
191622 c.3671C>A p.S1224Y Chr.17: 58571967-58571967 on Build GRCh38
5461415 c.3660G>T p.K1220N Chr.17: 58571978-58571978 on Build GRCh38
7116020 c.3824C>G p.S1275C Chr.17: 58571952-58571952 on Build GRCh38
95324 c.3568C>G p.Q1190E Chr.17: 58572070-58572070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755537604 Chr.17: 58572111-58572111 on Build GRCh38
rs756124299 Chr.17: 58571992-58571992 on Build GRCh38
rs576603353 Chr.17: 58571982-58571982 on Build GRCh38
rs372080024 Chr.17: 58571940-58571940 on Build GRCh38
rs138328740 Chr.17: 58571996-58571996 on Build GRCh38
rs774516557 Chr.17: 58571918-58571918 on Build GRCh38
rs571334539 Chr.17: 58571925-58571925 on Build GRCh38
rs759299104 Chr.17: 58572074-58572074 on Build GRCh38
rs374331123 Chr.17: 58572123-58572123 on Build GRCh38
rs748171773 Chr.17: 58571911-58571911 on Build GRCh38
rs192369207 Chr.17: 58571914-58571914 on Build GRCh38
rs766136939 Chr.17: 58571950-58571950 on Build GRCh38
rs149446135 Chr.17: 58572050-58572050 on Build GRCh38
rs747014221 Chr.17: 58572031-58572031 on Build GRCh38
rs746156882 Chr.17: 58572006-58572006 on Build GRCh38
rs753716897 Chr.17: 58572122-58572122 on Build GRCh38
rs150896699 Chr.17: 58571973-58571973 on Build GRCh38
rs936849107 Chr.17: 58571958-58571958 on Build GRCh38
rs780532737 Chr.17: 58572121-58572121 on Build GRCh38
rs772253260 Chr.17: 58571926-58571926 on Build GRCh38
rs770276375 Chr.17: 58572118-58572118 on Build GRCh38
rs775689003 Chr.17: 58572020-58572020 on Build GRCh38
rs761287596 Chr.17: 58571930-58571930 on Build GRCh38
rs553754986 Chr.17: 58572010-58572013 on Build GRCh38
rs948360612 Chr.17: 58572007-58572007 on Build GRCh38
rs200564361 Chr.17: 58571916-58571916 on Build GRCh38
rs769612454 Chr.17: 58571913-58571913 on Build GRCh38
rs772154999 Chr.17: 58572018-58572018 on Build GRCh38
rs775427906 Chr.17: 58571927-58571927 on Build GRCh38
rs764813019 Chr.17: 58571927-58571928 on Build GRCh38
rs778720258 Chr.17: 58572126-58572126 on Build GRCh38
rs761173950 Chr.17: 58571912-58571913 on Build GRCh38
rs975110914 Chr.17: 58571924-58571924 on Build GRCh38
rs923680431 Chr.17: 58571942-58571942 on Build GRCh38
rs771162846 Chr.17: 58571915-58571915 on Build GRCh38
rs187298992 Chr.17: 58571991-58571991 on Build GRCh38
rs773702832 Chr.17: 58572065-58572065 on Build GRCh38
rs541044627 Chr.17: 58572058-58572058 on Build GRCh38
rs201508314 Chr.17: 58571908-58571908 on Build GRCh38
rs752271471 Chr.17: 58572107-58572107 on Build GRCh38
rs767052427 Chr.17: 58572105-58572105 on Build GRCh38
rs906925347 Chr.17: 58572025-58572025 on Build GRCh38
rs756978985 Chr.17: 58571999-58571999 on Build GRCh38
rs781001403 Chr.17: 58571909-58571909 on Build GRCh38
rs184786723 Chr.17: 58571952-58571952 on Build GRCh38
rs370949525 Chr.17: 58572001-58572001 on Build GRCh38
rs769239541 Chr.17: 58572036-58572036 on Build GRCh38
rs141531297 Chr.17: 58571941-58571941 on Build GRCh38

View Full Product Details