Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 140686547-140686796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTK2
Entrez Gene ID
5747
Gene Name
protein tyrosine kinase 2
Gene Aliases
FADK, FAK, FAK1, FRNK, PPP1R71, p125FAK, pp125FAK
UniGene
Hs.395482
Gene Chromosome Location
Chr.8: 140658382-141001313 on Build GRCh38
Transcripts
NM_001199649.1 NM_001316342.1 NM_005607.4 NM_153831.3 XM_005251003.2 XM_005251012.2 XM_005251013.2 XM_006716608.2 XM_006716609.2 XM_006716610.2 XM_011517194.2 XM_017013651.1 XM_017013652.1 XM_017013653.1 XM_017013654.1 XM_017013655.1 XM_017013656.1 XM_017013657.1 XM_017013658.1 XM_017013659.1 XM_017013660.1 XM_017013661.1 XM_017013662.1 XM_017013663.1 XM_017013664.1 XM_017013665.1 XM_017013666.1 XM_017013667.1 XM_017013668.1 XM_017013669.1 XM_017013670.1 XM_017013671.1 XM_017013672.1 XM_017013673.1 XM_017013674.1 XM_017013675.1 XM_017013676.1 XM_017013677.1 XM_017013678.1 XM_017013679.1 XM_017013680.1 XM_017013681.1 XM_017013682.1 XM_017013683.1 XM_017013684.1 XM_017013685.1 XM_017013686.1 XM_017013687.1 XM_017013688.1 XM_017013689.1 XM_017013690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6758661 c.2417C>T p.P806L Chr.8: 140686633-140686633 on Build GRCh38
1455142 c.2377C>T p.R793* Chr.8: 140686673-140686673 on Build GRCh38
1455141 c.2521C>T p.R841* Chr.8: 140686673-140686673 on Build GRCh38
6758660 c.2561C>T p.P854L Chr.8: 140686633-140686633 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1037530100 Chr.8: 140686605-140686605 on Build GRCh38
rs745714715 Chr.8: 140686627-140686627 on Build GRCh38
rs762006575 Chr.8: 140686597-140686597 on Build GRCh38
rs768237414 Chr.8: 140686646-140686646 on Build GRCh38
rs773944025 Chr.8: 140686599-140686599 on Build GRCh38
rs769838913 Chr.8: 140686629-140686629 on Build GRCh38
rs773008171 Chr.8: 140686706-140686710 on Build GRCh38
rs897774876 Chr.8: 140686630-140686630 on Build GRCh38
rs745523477 Chr.8: 140686669-140686669 on Build GRCh38
rs773752021 Chr.8: 140686652-140686652 on Build GRCh38
rs760540866 Chr.8: 140686736-140686736 on Build GRCh38
rs879161446 Chr.8: 140686665-140686665 on Build GRCh38
rs775614348 Chr.8: 140686633-140686633 on Build GRCh38
rs374549965 Chr.8: 140686589-140686589 on Build GRCh38
rs778996100 Chr.8: 140686592-140686592 on Build GRCh38
rs377689571 Chr.8: 140686686-140686686 on Build GRCh38
rs748772540 Chr.8: 140686636-140686636 on Build GRCh38
rs1001847937 Chr.8: 140686687-140686687 on Build GRCh38
rs1021122229 Chr.8: 140686739-140686739 on Build GRCh38
rs755497817 Chr.8: 140686724-140686724 on Build GRCh38
rs780972917 Chr.8: 140686613-140686613 on Build GRCh38
rs200754638 Chr.8: 140686602-140686602 on Build GRCh38
rs761465770 Chr.8: 140686671-140686671 on Build GRCh38
rs146529989 Chr.8: 140686676-140686676 on Build GRCh38
rs41272409 Chr.8: 140686735-140686735 on Build GRCh38
rs771798292 Chr.8: 140686727-140686727 on Build GRCh38
rs773615646 Chr.8: 140686685-140686685 on Build GRCh38
rs199624251 Chr.8: 140686689-140686689 on Build GRCh38
rs754304372 Chr.8: 140686706-140686706 on Build GRCh38
rs767144547 Chr.8: 140686672-140686672 on Build GRCh38

View Full Product Details