Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1961740-1961965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFB10
Entrez Gene ID
4716
Gene Name
NADH:ubiquinone oxidoreductase subunit B10
Gene Aliases
PDSW
UniGene
Hs.513266
Gene Chromosome Location
Chr.16: 1959516-1961975 on Build GRCh38
Transcripts
NM_004548.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3741810 c.484A>C p.K162Q Chr.16: 1961871-1961871 on Build GRCh38
7233741 c.486A>G p.K162K Chr.16: 1961873-1961873 on Build GRCh38
1609006 c.446G>T p.C149F Chr.16: 1961833-1961833 on Build GRCh38
7193942 c.519A>C p.*173C Chr.16: 1961906-1961906 on Build GRCh38
1376305 c.461G>A p.R154K Chr.16: 1961848-1961848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567165467 Chr.16: 1961811-1961811 on Build GRCh38
rs765718162 Chr.16: 1961797-1961797 on Build GRCh38
rs765740331 Chr.16: 1961890-1961890 on Build GRCh38
rs745994055 Chr.16: 1961819-1961819 on Build GRCh38
rs763755867 Chr.16: 1961840-1961843 on Build GRCh38
rs766973448 Chr.16: 1961777-1961777 on Build GRCh38
rs765975212 Chr.16: 1961861-1961861 on Build GRCh38
rs757539908 Chr.16: 1961912-1961912 on Build GRCh38
rs747732434 Chr.16: 1961885-1961885 on Build GRCh38
rs141166307 Chr.16: 1961880-1961880 on Build GRCh38
rs201285379 Chr.16: 1961917-1961917 on Build GRCh38
rs775258391 Chr.16: 1961845-1961846 on Build GRCh38
rs150972831 Chr.16: 1961914-1961914 on Build GRCh38
rs923871086 Chr.16: 1961937-1961937 on Build GRCh38
rs751794350 Chr.16: 1961810-1961810 on Build GRCh38
rs371048972 Chr.16: 1961909-1961909 on Build GRCh38
rs779897644 Chr.16: 1961831-1961831 on Build GRCh38
rs201965625 Chr.16: 1961900-1961900 on Build GRCh38
rs749181367 Chr.16: 1961929-1961929 on Build GRCh38
rs780202758 Chr.16: 1961927-1961927 on Build GRCh38
rs1018333355 Chr.16: 1961866-1961866 on Build GRCh38
rs568374835 Chr.16: 1961823-1961823 on Build GRCh38
rs147766992 Chr.16: 1961812-1961812 on Build GRCh38
rs898194487 Chr.16: 1961839-1961839 on Build GRCh38
rs1008419869 Chr.16: 1961865-1961865 on Build GRCh38
rs375613670 Chr.16: 1961892-1961892 on Build GRCh38
rs372422108 Chr.16: 1961926-1961926 on Build GRCh38
rs145125673 Chr.16: 1961889-1961889 on Build GRCh38
rs1026346217 Chr.16: 1961845-1961845 on Build GRCh38
rs867416910 Chr.16: 1961834-1961834 on Build GRCh38
rs760026841 Chr.16: 1961796-1961796 on Build GRCh38
rs866543800 Chr.16: 1961875-1961875 on Build GRCh38
rs756074623 Chr.16: 1961827-1961827 on Build GRCh38
rs113497710 Chr.16: 1961896-1961896 on Build GRCh38
rs74903026 Chr.16: 1961924-1961924 on Build GRCh38
rs202033778 Chr.16: 1961766-1961766 on Build GRCh38
rs774039192 Chr.16: 1961765-1961765 on Build GRCh38
rs534213425 Chr.16: 1961888-1961888 on Build GRCh38
rs373969933 Chr.16: 1961788-1961788 on Build GRCh38
rs373787740 Chr.16: 1961805-1961805 on Build GRCh38
rs923778298 Chr.16: 1961772-1961772 on Build GRCh38
rs749906144 Chr.16: 1961784-1961784 on Build GRCh38
rs994412715 Chr.16: 1961840-1961840 on Build GRCh38
rs919014616 Chr.16: 1961895-1961895 on Build GRCh38
rs199975537 Chr.16: 1961894-1961894 on Build GRCh38
rs963150022 Chr.16: 1961891-1961891 on Build GRCh38
rs78924571 Chr.16: 1961873-1961873 on Build GRCh38
rs753098958 Chr.16: 1961804-1961804 on Build GRCh38
rs751990038 Chr.16: 1961901-1961901 on Build GRCh38
rs778306897 Chr.16: 1961806-1961806 on Build GRCh38

View Full Product Details