Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156134926-156135169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNA
Entrez Gene ID
4000
Gene Name
lamin A/C
Gene Aliases
CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, FPLD2, HGPS, IDC, LDP1, LFP, LGMD1B, LMN1, LMNC, LMNL1, PRO1
UniGene
Hs.594444
Gene Chromosome Location
Chr.1: 156082546-156140089 on Build GRCh38
Transcripts
NM_001257374.2 NM_001282624.1 NM_001282625.1 NM_001282626.1 NM_005572.3 NM_170707.3 NM_170708.3 XM_011509533.1 XM_011509534.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5495573 c.552G>A p.K184K Chr.1: 156134960-156134960 on Build GRCh38
7003933 c.800A>G p.Y267C Chr.1: 156134965-156134965 on Build GRCh38
5495572 c.795G>A p.K265K Chr.1: 156134960-156134960 on Build GRCh38
75247 c.767T>G p.V256G Chr.1: 156134932-156134932 on Build GRCh38
7003934 c.557A>G p.Y186C Chr.1: 156134965-156134965 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs397517909 Chr.1: 156134949-156134949 on Build GRCh38
rs267607602 Chr.1: 156135038-156135038 on Build GRCh38
rs776015136 Chr.1: 156135020-156135020 on Build GRCh38
rs267607625 Chr.1: 156134953-156134953 on Build GRCh38
rs949943976 Chr.1: 156134982-156134982 on Build GRCh38
rs774555415 Chr.1: 156135004-156135004 on Build GRCh38
rs267607630 Chr.1: 156134967-156134967 on Build GRCh38
rs749268854 Chr.1: 156134990-156134990 on Build GRCh38
rs556408262 Chr.1: 156135086-156135086 on Build GRCh38
rs750246389 Chr.1: 156134952-156134952 on Build GRCh38
rs267607632 Chr.1: 156134976-156134976 on Build GRCh38
rs746914371 Chr.1: 156134987-156134987 on Build GRCh38
rs771192755 Chr.1: 156134992-156134992 on Build GRCh38
rs1047858138 Chr.1: 156135073-156135073 on Build GRCh38
rs767732027 Chr.1: 156135119-156135119 on Build GRCh38
rs1009130231 Chr.1: 156135128-156135128 on Build GRCh38
rs746292194 Chr.1: 156135007-156135007 on Build GRCh38
rs772405351 Chr.1: 156135011-156135011 on Build GRCh38
rs768992495 Chr.1: 156135144-156135144 on Build GRCh38
rs879253920 Chr.1: 156134973-156134973 on Build GRCh38
rs980009386 Chr.1: 156134983-156134983 on Build GRCh38
rs267607593 Chr.1: 156134964-156134964 on Build GRCh38
rs886041211 Chr.1: 156134949-156134949 on Build GRCh38
rs746355819 Chr.1: 156135142-156135142 on Build GRCh38
rs907937372 Chr.1: 156135070-156135070 on Build GRCh38
rs148557956 Chr.1: 156134954-156134954 on Build GRCh38
rs780196737 Chr.1: 156134979-156134979 on Build GRCh38
rs267607631 Chr.1: 156134975-156134975 on Build GRCh38
rs75288240 Chr.1: 156135032-156135032 on Build GRCh38
rs11264444 Chr.1: 156134988-156134988 on Build GRCh38
rs1037561223 Chr.1: 156135116-156135116 on Build GRCh38
rs552365788 Chr.1: 156135025-156135025 on Build GRCh38
rs11264445 Chr.1: 156135036-156135036 on Build GRCh38
rs929457863 Chr.1: 156135057-156135057 on Build GRCh38
rs57048196 Chr.1: 156134965-156134965 on Build GRCh38
rs574775364 Chr.1: 156135123-156135123 on Build GRCh38

View Full Product Details