Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107923810-107924047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAMB1
Entrez Gene ID
3912
Gene Name
laminin subunit beta 1
Gene Aliases
CLM, LIS5
UniGene
Hs.650585
Gene Chromosome Location
Chr.7: 107923799-108003359 on Build GRCh38
Transcripts
NM_002291.2 XM_017012201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4697308 c.5301A>C p.E1767D Chr.7: 107924011-107924011 on Build GRCh38
3631815 c.5297G>A p.G1766E Chr.7: 107924015-107924015 on Build GRCh38
3877070 c.5336T>G p.V1779G Chr.7: 107923976-107923976 on Build GRCh38
252575 c.5306G>A p.R1769H Chr.7: 107924006-107924006 on Build GRCh38
5525176 c.5312T>A p.L1771H Chr.7: 107924000-107924000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911744619 Chr.7: 107923940-107923940 on Build GRCh38
rs374245297 Chr.7: 107923965-107923965 on Build GRCh38
rs777824210 Chr.7: 107924011-107924011 on Build GRCh38
rs770117339 Chr.7: 107924007-107924007 on Build GRCh38
rs781629439 Chr.7: 107923998-107923998 on Build GRCh38
rs368697174 Chr.7: 107923913-107923913 on Build GRCh38
rs571780695 Chr.7: 107923860-107923860 on Build GRCh38
rs138333734 Chr.7: 107923980-107923980 on Build GRCh38
rs753456642 Chr.7: 107923971-107923971 on Build GRCh38
rs775157505 Chr.7: 107923918-107923918 on Build GRCh38
rs767997731 Chr.7: 107923964-107923964 on Build GRCh38
rs756798281 Chr.7: 107923976-107923976 on Build GRCh38
rs769346376 Chr.7: 107923943-107923945 on Build GRCh38
rs748405251 Chr.7: 107923917-107923917 on Build GRCh38
rs369886731 Chr.7: 107923954-107923954 on Build GRCh38
rs748443031 Chr.7: 107924006-107924006 on Build GRCh38
rs755468191 Chr.7: 107923993-107923993 on Build GRCh38
rs7561 Chr.7: 107923921-107923921 on Build GRCh38
rs1033829868 Chr.7: 107923877-107923877 on Build GRCh38
rs774935596 Chr.7: 107923964-107923965 on Build GRCh38
rs369528849 Chr.7: 107923987-107923987 on Build GRCh38
rs573998500 Chr.7: 107923950-107923950 on Build GRCh38
rs763641113 Chr.7: 107923966-107923966 on Build GRCh38
rs959366589 Chr.7: 107923892-107923892 on Build GRCh38
rs745460957 Chr.7: 107923939-107923940 on Build GRCh38
rs762934032 Chr.7: 107923919-107923919 on Build GRCh38
rs977935801 Chr.7: 107923934-107923934 on Build GRCh38
rs78245660 Chr.7: 107923866-107923866 on Build GRCh38
rs944465476 Chr.7: 107923949-107923949 on Build GRCh38

View Full Product Details