Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64254911-64255164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLCB3
Entrez Gene ID
5331
Gene Name
phospholipase C beta 3
Gene Aliases
-
UniGene
Hs.523761
Gene Chromosome Location
Chr.11: 64251523-64269452 on Build GRCh38
Transcripts
NM_000932.2 NM_001184883.1 NM_001316314.1 XM_011545101.2 XM_017017925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6519207 c.375T>A p.D125E Chr.11: 64255026-64255026 on Build GRCh38
689741 c.298G>T p.E100* Chr.11: 64254949-64254949 on Build GRCh38
930044 c.366C>T p.A122A Chr.11: 64255017-64255017 on Build GRCh38
5592564 c.284C>T p.P95L Chr.11: 64254935-64254935 on Build GRCh38
1676136 c.335C>T p.P112L Chr.11: 64254986-64254986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552167557 Chr.11: 64254981-64254981 on Build GRCh38
rs756426279 Chr.11: 64255039-64255040 on Build GRCh38
rs772667527 Chr.11: 64255012-64255012 on Build GRCh38
rs143389743 Chr.11: 64254932-64254932 on Build GRCh38
rs555033217 Chr.11: 64255096-64255096 on Build GRCh38
rs568976556 Chr.11: 64255114-64255114 on Build GRCh38
rs769130781 Chr.11: 64255018-64255018 on Build GRCh38
rs540937471 Chr.11: 64254965-64254965 on Build GRCh38
rs758373860 Chr.11: 64254937-64254937 on Build GRCh38
rs1040835588 Chr.11: 64255092-64255092 on Build GRCh38
rs11231731 Chr.11: 64254958-64254958 on Build GRCh38
rs868568800 Chr.11: 64255038-64255039 on Build GRCh38
rs764831153 Chr.11: 64255061-64255061 on Build GRCh38
rs750770829 Chr.11: 64255059-64255059 on Build GRCh38
rs752205723 Chr.11: 64255074-64255074 on Build GRCh38
rs752156172 Chr.11: 64254936-64254936 on Build GRCh38
rs769921677 Chr.11: 64254986-64254986 on Build GRCh38
rs780364862 Chr.11: 64255083-64255103 on Build GRCh38
rs373884767 Chr.11: 64255029-64255029 on Build GRCh38
rs762160886 Chr.11: 64255034-64255034 on Build GRCh38
rs750777504 Chr.11: 64254948-64254950 on Build GRCh38
rs777872410 Chr.11: 64254943-64254943 on Build GRCh38
rs1042935815 Chr.11: 64255095-64255095 on Build GRCh38
rs930971905 Chr.11: 64254969-64254969 on Build GRCh38
rs759166003 Chr.11: 64255060-64255060 on Build GRCh38
rs632439 Chr.11: 64254987-64254987 on Build GRCh38
rs1056763317 Chr.11: 64255106-64255106 on Build GRCh38
rs758028143 Chr.11: 64255086-64255086 on Build GRCh38
rs746158762 Chr.11: 64254966-64254966 on Build GRCh38
rs191435522 Chr.11: 64255105-64255105 on Build GRCh38
rs528884648 Chr.11: 64255044-64255044 on Build GRCh38
rs746775656 Chr.11: 64254944-64254944 on Build GRCh38

View Full Product Details