Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25362416-25362641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHCE
Entrez Gene ID
6006
Gene Name
Rh blood group CcEe antigens
Gene Aliases
CD240CE, RH, RH30A, RHC, RHE, RHIXB, RHPI, Rh4, RhIVb(J), RhVI, RhVIII
UniGene
Hs.449968
Gene Chromosome Location
Chr.1: 25362249-25430193 on Build GRCh38
Transcripts
NM_020485.4 NM_138616.3 NM_138617.3 NM_138618.3 XM_005245957.3 XM_011541888.2 XM_011541889.2 XM_011541890.2 XM_011541891.2 XM_017002014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5195398 c.1176T>G p.R392R Chr.1: 25362451-25362451 on Build GRCh38
6317182 c.1171G>T p.V391L Chr.1: 25362456-25362456 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779686528 Chr.1: 25362516-25362517 on Build GRCh38
rs752872977 Chr.1: 25362589-25362589 on Build GRCh38
rs771897306 Chr.1: 25362494-25362494 on Build GRCh38
rs781442079 Chr.1: 25362449-25362449 on Build GRCh38
rs945863301 Chr.1: 25362611-25362611 on Build GRCh38
rs755024990 Chr.1: 25362444-25362444 on Build GRCh38
rs766663148 Chr.1: 25362561-25362561 on Build GRCh38
rs781386809 Chr.1: 25362578-25362578 on Build GRCh38
rs748337019 Chr.1: 25362453-25362453 on Build GRCh38
rs750786353 Chr.1: 25362556-25362556 on Build GRCh38
rs751180355 Chr.1: 25362555-25362556 on Build GRCh38
rs988186229 Chr.1: 25362550-25362550 on Build GRCh38
rs770177192 Chr.1: 25362456-25362456 on Build GRCh38
rs776158672 Chr.1: 25362523-25362523 on Build GRCh38
rs752015325 Chr.1: 25362443-25362443 on Build GRCh38
rs376075232 Chr.1: 25362498-25362498 on Build GRCh38
rs755467449 Chr.1: 25362568-25362568 on Build GRCh38
rs914028916 Chr.1: 25362609-25362609 on Build GRCh38
rs763314236 Chr.1: 25362560-25362560 on Build GRCh38
rs775429475 Chr.1: 25362495-25362495 on Build GRCh38
rs764930503 Chr.1: 25362534-25362534 on Build GRCh38
rs754715308 Chr.1: 25362596-25362597 on Build GRCh38
rs778828022 Chr.1: 25362458-25362458 on Build GRCh38
rs752052778 Chr.1: 25362563-25362563 on Build GRCh38
rs758007822 Chr.1: 25362490-25362490 on Build GRCh38
rs745715622 Chr.1: 25362462-25362462 on Build GRCh38
rs768174478 Chr.1: 25362502-25362502 on Build GRCh38
rs761433940 Chr.1: 25362532-25362532 on Build GRCh38

View Full Product Details