Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 57633849-57634110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDH17
Entrez Gene ID
27253
Gene Name
protocadherin 17
Gene Aliases
PCDH68, PCH68
UniGene
Hs.106511
Gene Chromosome Location
Chr.13: 57630104-57728931 on Build GRCh38
Transcripts
NM_001040429.2 XM_005266357.2 XM_005266358.2 XM_017020547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5362047 c.1339delG p.G448fs*16 Chr.13: 57633885-57633885 on Build GRCh38
948160 c.1407G>A p.P469P Chr.13: 57633953-57633953 on Build GRCh38
5342371 c.1514G>A p.G505D Chr.13: 57634060-57634060 on Build GRCh38
5362201 c.1446C>T p.H482H Chr.13: 57633992-57633992 on Build GRCh38
1219469 c.1500C>A p.D500E Chr.13: 57634046-57634046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575099150 Chr.13: 57633971-57633971 on Build GRCh38
rs756962330 Chr.13: 57634075-57634075 on Build GRCh38
rs763890261 Chr.13: 57633900-57633900 on Build GRCh38
rs958427962 Chr.13: 57634026-57634026 on Build GRCh38
rs1026845535 Chr.13: 57633917-57633917 on Build GRCh38
rs369545682 Chr.13: 57633926-57633926 on Build GRCh38
rs752772444 Chr.13: 57634025-57634025 on Build GRCh38
rs758916766 Chr.13: 57633886-57633886 on Build GRCh38
rs371111071 Chr.13: 57634064-57634064 on Build GRCh38
rs201009234 Chr.13: 57633964-57633964 on Build GRCh38
rs779055194 Chr.13: 57633870-57633870 on Build GRCh38
rs758609525 Chr.13: 57633933-57633933 on Build GRCh38
rs768355749 Chr.13: 57633981-57633981 on Build GRCh38
rs868545491 Chr.13: 57633963-57633963 on Build GRCh38
rs758480081 Chr.13: 57634042-57634042 on Build GRCh38
rs539857227 Chr.13: 57633918-57633918 on Build GRCh38
rs762665985 Chr.13: 57633899-57633899 on Build GRCh38
rs770772846 Chr.13: 57633885-57633885 on Build GRCh38
rs755006939 Chr.13: 57633922-57633922 on Build GRCh38
rs761103875 Chr.13: 57633908-57633908 on Build GRCh38
rs766794348 Chr.13: 57633992-57633992 on Build GRCh38
rs976333418 Chr.13: 57634088-57634088 on Build GRCh38
rs772965892 Chr.13: 57633880-57633880 on Build GRCh38
rs368442905 Chr.13: 57633957-57633957 on Build GRCh38
rs760899233 Chr.13: 57633986-57633986 on Build GRCh38
rs138615233 Chr.13: 57634047-57634047 on Build GRCh38
rs145861936 Chr.13: 57633872-57633872 on Build GRCh38
rs569566282 Chr.13: 57633904-57633904 on Build GRCh38
rs376127001 Chr.13: 57633887-57633887 on Build GRCh38
rs765788321 Chr.13: 57634021-57634021 on Build GRCh38
rs777803892 Chr.13: 57634046-57634046 on Build GRCh38
rs754390922 Chr.13: 57633921-57633921 on Build GRCh38
rs746719630 Chr.13: 57633942-57633942 on Build GRCh38
rs866861544 Chr.13: 57633962-57633962 on Build GRCh38
rs1020036058 Chr.13: 57633935-57633935 on Build GRCh38
rs562352047 Chr.13: 57634013-57634013 on Build GRCh38
rs374629436 Chr.13: 57633938-57633938 on Build GRCh38
rs760011678 Chr.13: 57634016-57634016 on Build GRCh38
rs781010005 Chr.13: 57634077-57634077 on Build GRCh38
rs774066378 Chr.13: 57633983-57633983 on Build GRCh38
rs772318001 Chr.13: 57633876-57633876 on Build GRCh38
rs41292836 Chr.13: 57633959-57633959 on Build GRCh38
rs745707564 Chr.13: 57634080-57634080 on Build GRCh38
rs778930576 Chr.13: 57633925-57633925 on Build GRCh38
rs775243804 Chr.13: 57633897-57633897 on Build GRCh38
rs889733374 Chr.13: 57633920-57633920 on Build GRCh38
rs769843004 Chr.13: 57634085-57634085 on Build GRCh38
rs531156670 Chr.13: 57634091-57634091 on Build GRCh38
rs548939475 Chr.13: 57634007-57634007 on Build GRCh38
rs1056463297 Chr.13: 57633871-57633871 on Build GRCh38
rs368374245 Chr.13: 57633884-57633884 on Build GRCh38
rs770777439 Chr.13: 57633953-57633953 on Build GRCh38
rs746882471 Chr.13: 57633881-57633881 on Build GRCh38

View Full Product Details