Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102423624-102423897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18RAP
Entrez Gene ID
8807
Gene Name
interleukin 18 receptor accessory protein
Gene Aliases
ACPL, CD218b, CDw218b, IL-18R-beta, IL-18RAcP, IL-18Rbeta, IL-1R-7, IL-1R7, IL-1RAcPL, IL18RB
UniGene
Hs.158315
Gene Chromosome Location
Chr.2: 102418551-102452568 on Build GRCh38
Transcripts
NM_003853.3 XM_011512087.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1398525 c.81delA p.K29fs*66 Chr.2: 102423821-102423821 on Build GRCh38
4523555 c.121G>A p.E41K Chr.2: 102423861-102423861 on Build GRCh38
4759602 c.127G>A p.V43I Chr.2: 102423867-102423867 on Build GRCh38
7276701 c.80_81delCA p.T27fs*12 Chr.2: 102423820-102423821 on Build GRCh38
4893235 c.139G>A p.D47N Chr.2: 102423879-102423879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753851163 Chr.2: 102423857-102423857 on Build GRCh38
rs762023114 Chr.2: 102423823-102423823 on Build GRCh38
rs756319084 Chr.2: 102423821-102423821 on Build GRCh38
rs371728977 Chr.2: 102423801-102423801 on Build GRCh38
rs6759588 Chr.2: 102423699-102423699 on Build GRCh38
rs978389263 Chr.2: 102423648-102423648 on Build GRCh38
rs936895996 Chr.2: 102423677-102423677 on Build GRCh38
rs751866033 Chr.2: 102423785-102423786 on Build GRCh38
rs767626133 Chr.2: 102423827-102423827 on Build GRCh38
rs3215320 Chr.2: 102423792-102423793 on Build GRCh38
rs369898608 Chr.2: 102423867-102423867 on Build GRCh38
rs924088939 Chr.2: 102423664-102423664 on Build GRCh38
rs750538918 Chr.2: 102423837-102423837 on Build GRCh38
rs774512653 Chr.2: 102423815-102423815 on Build GRCh38
rs1026827083 Chr.2: 102423814-102423814 on Build GRCh38
rs869042654 Chr.2: 102423793-102423794 on Build GRCh38
rs9807934 Chr.2: 102423722-102423722 on Build GRCh38
rs770263971 Chr.2: 102423785-102423785 on Build GRCh38
rs777152543 Chr.2: 102423774-102423774 on Build GRCh38
rs11377261 Chr.2: 102423787-102423788 on Build GRCh38
rs1028469626 Chr.2: 102423778-102423778 on Build GRCh38
rs201039117 Chr.2: 102423789-102423790 on Build GRCh38
rs760633536 Chr.2: 102423850-102423850 on Build GRCh38
rs376556225 Chr.2: 102423789-102423789 on Build GRCh38
rs746383375 Chr.2: 102423776-102423776 on Build GRCh38
rs149062947 Chr.2: 102423695-102423695 on Build GRCh38
rs1047891368 Chr.2: 102423705-102423705 on Build GRCh38
rs2272129 Chr.2: 102423789-102423789 on Build GRCh38
rs1003190722 Chr.2: 102423719-102423719 on Build GRCh38
rs138870268 Chr.2: 102423689-102423689 on Build GRCh38
rs6543133 Chr.2: 102423717-102423717 on Build GRCh38
rs752896941 Chr.2: 102423791-102423793 on Build GRCh38
rs11465689 Chr.2: 102423707-102423707 on Build GRCh38
rs398104566 Chr.2: 102423788-102423789 on Build GRCh38
rs1044017953 Chr.2: 102423710-102423710 on Build GRCh38
rs766278345 Chr.2: 102423851-102423851 on Build GRCh38
rs143062976 Chr.2: 102423721-102423721 on Build GRCh38

View Full Product Details