Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 222624649-222624898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIA3
Entrez Gene ID
375056
Gene Name
MIA family member 3, ER export factor
Gene Aliases
ARNT, D320, TANGO, TANGO1, UNQ6077
UniGene
Hs.118474
Gene Chromosome Location
Chr.1: 222617790-222668012 on Build GRCh38
Transcripts
NM_001324062.1 NM_001324063.1 NM_001324064.1 NM_198551.3 XM_006711304.3 XM_011509513.2 XM_017001243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4485411 c.292C>T p.P98S Chr.1: 222624792-222624792 on Build GRCh38
904287 c.316C>A p.H106N Chr.1: 222624816-222624816 on Build GRCh38
1959864 c.293C>T p.P98L Chr.1: 222624793-222624793 on Build GRCh38
4702072 c.298G>T p.D100Y Chr.1: 222624798-222624798 on Build GRCh38
321686 c.354+2T>C p.? Chr.1: 222624856-222624856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758480502 Chr.1: 222624754-222624754 on Build GRCh38
rs555852727 Chr.1: 222624757-222624757 on Build GRCh38
rs775032876 Chr.1: 222624706-222624706 on Build GRCh38
rs375571811 Chr.1: 222624753-222624753 on Build GRCh38
rs763708048 Chr.1: 222624775-222624775 on Build GRCh38
rs758603021 Chr.1: 222624758-222624758 on Build GRCh38
rs777860336 Chr.1: 222624758-222624758 on Build GRCh38
rs372980758 Chr.1: 222624774-222624774 on Build GRCh38
rs755184494 Chr.1: 222624752-222624752 on Build GRCh38
rs766384095 Chr.1: 222624718-222624718 on Build GRCh38
rs573713766 Chr.1: 222624870-222624870 on Build GRCh38
rs922405504 Chr.1: 222624755-222624755 on Build GRCh38
rs748507677 Chr.1: 222624860-222624860 on Build GRCh38
rs769464287 Chr.1: 222624820-222624820 on Build GRCh38
rs555552594 Chr.1: 222624869-222624869 on Build GRCh38
rs3008617 Chr.1: 222624764-222624764 on Build GRCh38
rs903715700 Chr.1: 222624681-222624681 on Build GRCh38
rs553838790 Chr.1: 222624826-222624826 on Build GRCh38
rs374726806 Chr.1: 222624808-222624808 on Build GRCh38
rs551297427 Chr.1: 222624705-222624705 on Build GRCh38
rs746885908 Chr.1: 222624760-222624760 on Build GRCh38
rs986735282 Chr.1: 222624747-222624747 on Build GRCh38
rs376408958 Chr.1: 222624748-222624748 on Build GRCh38
rs956871695 Chr.1: 222624710-222624710 on Build GRCh38

View Full Product Details