Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226064315-226064576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H3F3A
Entrez Gene ID
3020
Gene Name
H3 histone, family 3A
Gene Aliases
H3.3A, H3F3
UniGene
Hs.533624
Gene Chromosome Location
Chr.1: 226062707-226072002 on Build GRCh38
Transcripts
NM_002107.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5024960 c.18G>C p.Q6H Chr.1: 226064369-226064369 on Build GRCh38
4971457 c.81C>T p.R27R Chr.1: 226064432-226064432 on Build GRCh38
1578107 c.84G>T p.K28N Chr.1: 226064435-226064435 on Build GRCh38
5574356 c.107_108insTCT p.V36_K37insL Chr.1: 226064458-226064459 on Build GRCh38
123523 c.7C>T p.R3C Chr.1: 226064358-226064358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888157382 Chr.1: 226064435-226064435 on Build GRCh38
rs754254437 Chr.1: 226064341-226064342 on Build GRCh38
rs763924881 Chr.1: 226064528-226064528 on Build GRCh38
rs765706112 Chr.1: 226064485-226064485 on Build GRCh38
rs546859287 Chr.1: 226064508-226064509 on Build GRCh38
rs2036870 Chr.1: 226064531-226064531 on Build GRCh38
rs762415821 Chr.1: 226064484-226064485 on Build GRCh38
rs751837333 Chr.1: 226064512-226064513 on Build GRCh38
rs759646833 Chr.1: 226064494-226064494 on Build GRCh38
rs758609953 Chr.1: 226064494-226064495 on Build GRCh38
rs750509756 Chr.1: 226064485-226064488 on Build GRCh38
rs773440544 Chr.1: 226064399-226064399 on Build GRCh38
rs762679777 Chr.1: 226064513-226064513 on Build GRCh38
rs1017424794 Chr.1: 226064384-226064384 on Build GRCh38
rs200633260 Chr.1: 226064429-226064429 on Build GRCh38
rs1032345596 Chr.1: 226064456-226064456 on Build GRCh38
rs1021695176 Chr.1: 226064501-226064501 on Build GRCh38
rs747176447 Chr.1: 226064420-226064420 on Build GRCh38
rs772383036 Chr.1: 226064393-226064393 on Build GRCh38
rs376524805 Chr.1: 226064502-226064502 on Build GRCh38
rs370230636 Chr.1: 226064495-226064495 on Build GRCh38
rs755815078 Chr.1: 226064518-226064519 on Build GRCh38
rs748498139 Chr.1: 226064363-226064363 on Build GRCh38
rs776640745 Chr.1: 226064465-226064465 on Build GRCh38
rs199699563 Chr.1: 226064346-226064346 on Build GRCh38

View Full Product Details