Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48527755-48527990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8S1
Entrez Gene ID
341568
Gene Name
olfactory receptor family 8 subfamily S member 1
Gene Aliases
-
UniGene
Hs.524373
Gene Chromosome Location
Chr.12: 48525632-48528103 on Build GRCh38
Transcripts
NM_001005203.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2005967 c.923C>G p.P308R Chr.12: 48527946-48527946 on Build GRCh38
2005968 c.950G>A p.W317* Chr.12: 48527973-48527973 on Build GRCh38
6321549 c.943G>T p.A315S Chr.12: 48527966-48527966 on Build GRCh38
2005966 c.905G>A p.R302Q Chr.12: 48527928-48527928 on Build GRCh38
5042602 c.958C>T p.P320S Chr.12: 48527981-48527981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750486295 Chr.12: 48527889-48527890 on Build GRCh38
rs201251512 Chr.12: 48527898-48527898 on Build GRCh38
rs759222823 Chr.12: 48527928-48527928 on Build GRCh38
rs775798972 Chr.12: 48527896-48527896 on Build GRCh38
rs768162058 Chr.12: 48527908-48527908 on Build GRCh38
rs200743626 Chr.12: 48527900-48527900 on Build GRCh38
rs368455271 Chr.12: 48527870-48527870 on Build GRCh38
rs372740627 Chr.12: 48527881-48527881 on Build GRCh38
rs777773697 Chr.12: 48527966-48527966 on Build GRCh38
rs765508225 Chr.12: 48527862-48527865 on Build GRCh38
rs539389734 Chr.12: 48527879-48527879 on Build GRCh38
rs772527904 Chr.12: 48527913-48527913 on Build GRCh38
rs749535200 Chr.12: 48527897-48527897 on Build GRCh38
rs1049478599 Chr.12: 48527869-48527869 on Build GRCh38
rs575806552 Chr.12: 48527967-48527967 on Build GRCh38
rs758417729 Chr.12: 48527898-48527899 on Build GRCh38
rs764990731 Chr.12: 48527929-48527929 on Build GRCh38
rs762388664 Chr.12: 48527902-48527902 on Build GRCh38
rs778051348 Chr.12: 48527934-48527934 on Build GRCh38
rs914053386 Chr.12: 48527865-48527865 on Build GRCh38
rs758289378 Chr.12: 48527965-48527965 on Build GRCh38
rs769844098 Chr.12: 48527888-48527888 on Build GRCh38
rs758376787 Chr.12: 48527866-48527866 on Build GRCh38
rs752508783 Chr.12: 48527941-48527941 on Build GRCh38
rs115731024 Chr.12: 48527868-48527868 on Build GRCh38
rs536266296 Chr.12: 48527827-48527827 on Build GRCh38
rs765862542 Chr.12: 48527921-48527921 on Build GRCh38
rs753524189 Chr.12: 48527927-48527927 on Build GRCh38
rs567121917 Chr.12: 48527792-48527792 on Build GRCh38
rs527798750 Chr.12: 48527858-48527858 on Build GRCh38
rs760245164 Chr.12: 48527919-48527919 on Build GRCh38

View Full Product Details