Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136825135-136825390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX7
Entrez Gene ID
5191
Gene Name
peroxisomal biogenesis factor 7
Gene Aliases
PBD9B, PTS2R, RCDP1, RD
UniGene
Hs.280932
Gene Chromosome Location
Chr.6: 136822220-136913934 on Build GRCh38
Transcripts
NM_000288.3 XM_005267019.4 XM_006715502.1 XM_017010934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6868350 c.172C>T p.L58L Chr.6: 136825255-136825255 on Build GRCh38
6339828 c.131-8C>T p.? Chr.6: 136825206-136825206 on Build GRCh38
6868349 c.172C>T p.L58L Chr.6: 136825255-136825255 on Build GRCh38
6339829 c.131-8C>T p.? Chr.6: 136825206-136825206 on Build GRCh38
5547991 c.188+5delG p.? Chr.6: 136825276-136825276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774131564 Chr.6: 136825262-136825262 on Build GRCh38
rs781249811 Chr.6: 136825295-136825295 on Build GRCh38
rs772980172 Chr.6: 136825192-136825192 on Build GRCh38
rs774015364 Chr.6: 136825215-136825215 on Build GRCh38
rs761605500 Chr.6: 136825261-136825262 on Build GRCh38
rs773174452 Chr.6: 136825309-136825309 on Build GRCh38
rs200234391 Chr.6: 136825274-136825274 on Build GRCh38
rs1025379862 Chr.6: 136825362-136825362 on Build GRCh38
rs767077870 Chr.6: 136825242-136825242 on Build GRCh38
rs995456495 Chr.6: 136825300-136825300 on Build GRCh38
rs566604333 Chr.6: 136825294-136825294 on Build GRCh38
rs752812577 Chr.6: 136825298-136825298 on Build GRCh38
rs765917993 Chr.6: 136825206-136825206 on Build GRCh38
rs376679254 Chr.6: 136825244-136825244 on Build GRCh38
rs765902458 Chr.6: 136825250-136825250 on Build GRCh38
rs760985736 Chr.6: 136825197-136825198 on Build GRCh38
rs747201722 Chr.6: 136825290-136825290 on Build GRCh38
rs758835413 Chr.6: 136825267-136825267 on Build GRCh38
rs750791932 Chr.6: 136825254-136825254 on Build GRCh38
rs946227598 Chr.6: 136825198-136825198 on Build GRCh38
rs372505409 Chr.6: 136825174-136825174 on Build GRCh38
rs748015289 Chr.6: 136825180-136825180 on Build GRCh38
rs558638509 Chr.6: 136825368-136825368 on Build GRCh38
rs758230233 Chr.6: 136825255-136825255 on Build GRCh38
rs759134071 Chr.6: 136825225-136825225 on Build GRCh38
rs748753720 Chr.6: 136825205-136825206 on Build GRCh38
rs769643042 Chr.6: 136825198-136825198 on Build GRCh38
rs769793590 Chr.6: 136825190-136825190 on Build GRCh38
rs749215714 Chr.6: 136825311-136825311 on Build GRCh38
rs769699954 Chr.6: 136825302-136825302 on Build GRCh38
rs375687969 Chr.6: 136825268-136825268 on Build GRCh38
rs1048818118 Chr.6: 136825317-136825317 on Build GRCh38
rs373960146 Chr.6: 136825178-136825178 on Build GRCh38
rs757833805 Chr.6: 136825249-136825249 on Build GRCh38
rs267608254 Chr.6: 136825272-136825272 on Build GRCh38
rs370604445 Chr.6: 136825296-136825296 on Build GRCh38
rs770711981 Chr.6: 136825313-136825313 on Build GRCh38
rs768349731 Chr.6: 136825209-136825210 on Build GRCh38
rs768736911 Chr.6: 136825165-136825165 on Build GRCh38
rs762857800 Chr.6: 136825196-136825196 on Build GRCh38

View Full Product Details