Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5623953-5624224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF216
Entrez Gene ID
54476
Gene Name
ring finger protein 216
Gene Aliases
CAHH, TRIAD3, U7I1, UBCE7IP1, ZIN
UniGene
Hs.487458
Gene Chromosome Location
Chr.7: 5620041-5781730 on Build GRCh38
Transcripts
NM_207111.3 NM_207116.2 XM_005249785.2 XM_011515436.1 XM_017012363.1 XM_017012365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
421904 c.2425G>A p.E809K Chr.7: 5624083-5624083 on Build GRCh38
4833366 c.2445G>A p.K815K Chr.7: 5624063-5624063 on Build GRCh38
7286526 c.2383-7G>A p.? Chr.7: 5624132-5624132 on Build GRCh38
421905 c.2445G>T p.K815N Chr.7: 5624063-5624063 on Build GRCh38
4435556 c.2389G>C p.D797H Chr.7: 5624119-5624119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558323779 Chr.7: 5624015-5624015 on Build GRCh38
rs759265957 Chr.7: 5624104-5624104 on Build GRCh38
rs953443193 Chr.7: 5624038-5624038 on Build GRCh38
rs765019115 Chr.7: 5624109-5624109 on Build GRCh38
rs770633469 Chr.7: 5624077-5624077 on Build GRCh38
rs748610411 Chr.7: 5624056-5624056 on Build GRCh38
rs894121226 Chr.7: 5624201-5624201 on Build GRCh38
rs934053482 Chr.7: 5624003-5624003 on Build GRCh38
rs934686219 Chr.7: 5624016-5624016 on Build GRCh38
rs369778446 Chr.7: 5624155-5624155 on Build GRCh38
rs760339546 Chr.7: 5624070-5624070 on Build GRCh38
rs372528580 Chr.7: 5624040-5624040 on Build GRCh38
rs756339470 Chr.7: 5624010-5624010 on Build GRCh38
rs772623870 Chr.7: 5624066-5624066 on Build GRCh38
rs573156641 Chr.7: 5623999-5623999 on Build GRCh38
rs762819460 Chr.7: 5624120-5624120 on Build GRCh38
rs909100022 Chr.7: 5624060-5624060 on Build GRCh38
rs377507486 Chr.7: 5624133-5624133 on Build GRCh38
rs373744722 Chr.7: 5624166-5624166 on Build GRCh38
rs540592129 Chr.7: 5624156-5624156 on Build GRCh38
rs752588814 Chr.7: 5624116-5624116 on Build GRCh38
rs755333407 Chr.7: 5624150-5624150 on Build GRCh38
rs746570618 Chr.7: 5624157-5624157 on Build GRCh38
rs921105165 Chr.7: 5623993-5623993 on Build GRCh38
rs907420661 Chr.7: 5624114-5624114 on Build GRCh38
rs776393751 Chr.7: 5624086-5624086 on Build GRCh38
rs941861619 Chr.7: 5624117-5624117 on Build GRCh38
rs559249805 Chr.7: 5624178-5624178 on Build GRCh38
rs879527283 Chr.7: 5624129-5624129 on Build GRCh38
rs766772595 Chr.7: 5624142-5624142 on Build GRCh38
rs368788748 Chr.7: 5624202-5624202 on Build GRCh38
rs975202067 Chr.7: 5623989-5623989 on Build GRCh38
rs372775923 Chr.7: 5624183-5624183 on Build GRCh38
rs769275094 Chr.7: 5624036-5624036 on Build GRCh38
rs868517013 Chr.7: 5624179-5624179 on Build GRCh38
rs779439899 Chr.7: 5624154-5624154 on Build GRCh38
rs756345098 Chr.7: 5624140-5624140 on Build GRCh38
rs767612061 Chr.7: 5624132-5624132 on Build GRCh38
rs370743255 Chr.7: 5624175-5624175 on Build GRCh38
rs368999318 Chr.7: 5624007-5624007 on Build GRCh38
rs780534871 Chr.7: 5624017-5624017 on Build GRCh38
rs752785925 Chr.7: 5624110-5624110 on Build GRCh38
rs377645168 Chr.7: 5624168-5624168 on Build GRCh38
rs551351181 Chr.7: 5624006-5624006 on Build GRCh38
rs1013129237 Chr.7: 5624048-5624048 on Build GRCh38
rs754115852 Chr.7: 5624148-5624148 on Build GRCh38
rs951178006 Chr.7: 5624182-5624182 on Build GRCh38
rs771574713 Chr.7: 5624058-5624058 on Build GRCh38
rs749739000 Chr.7: 5624024-5624024 on Build GRCh38

View Full Product Details