Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 236380506-236380747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCA1
Entrez Gene ID
79781
Gene Name
IQ motif containing with AAA domain 1
Gene Aliases
4930465P12Rik, DRC11, IQCA
UniGene
Hs.591594
Gene Chromosome Location
Chr.2: 236324147-236507558 on Build GRCh38
Transcripts
NM_001270584.1 NM_001270585.1 NM_024726.4 NR_073043.1 XM_011511864.1 XM_011511865.2 XM_011511866.1 XM_017004960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4436191 c.1467G>C p.L489L Chr.2: 236380580-236380580 on Build GRCh38
1210842 c.1414-3A>G p.? Chr.2: 236380636-236380636 on Build GRCh38
6498916 c.1422G>C p.K474N Chr.2: 236380625-236380625 on Build GRCh38
6569479 c.1414-9_1414>T p.? Chr.2: 236380633-236380642 on Build GRCh38
5074842 c.1447G>A p.A483T Chr.2: 236380600-236380600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775984343 Chr.2: 236380570-236380570 on Build GRCh38
rs999118763 Chr.2: 236380727-236380727 on Build GRCh38
rs531455437 Chr.2: 236380661-236380661 on Build GRCh38
rs751193001 Chr.2: 236380593-236380593 on Build GRCh38
rs1027463141 Chr.2: 236380549-236380551 on Build GRCh38
rs777195271 Chr.2: 236380553-236380553 on Build GRCh38
rs941190866 Chr.2: 236380557-236380557 on Build GRCh38
rs371969607 Chr.2: 236380556-236380556 on Build GRCh38
rs974472217 Chr.2: 236380622-236380624 on Build GRCh38
rs973418567 Chr.2: 236380547-236380547 on Build GRCh38
rs577099007 Chr.2: 236380537-236380537 on Build GRCh38
rs950524462 Chr.2: 236380549-236380549 on Build GRCh38
rs944620924 Chr.2: 236380601-236380601 on Build GRCh38
rs113436711 Chr.2: 236380713-236380713 on Build GRCh38
rs761736345 Chr.2: 236380608-236380608 on Build GRCh38
rs750420394 Chr.2: 236380702-236380702 on Build GRCh38
rs929908691 Chr.2: 236380539-236380539 on Build GRCh38
rs770923582 Chr.2: 236380705-236380705 on Build GRCh38
rs796364621 Chr.2: 236380601-236380603 on Build GRCh38
rs865786826 Chr.2: 236380598-236380598 on Build GRCh38
rs776910442 Chr.2: 236380591-236380599 on Build GRCh38
rs767376217 Chr.2: 236380695-236380695 on Build GRCh38
rs866719628 Chr.2: 236380619-236380619 on Build GRCh38
rs902815579 Chr.2: 236380717-236380717 on Build GRCh38
rs935741538 Chr.2: 236380664-236380664 on Build GRCh38
rs374961030 Chr.2: 236380640-236380640 on Build GRCh38
rs899562588 Chr.2: 236380687-236380687 on Build GRCh38
rs77528566 Chr.2: 236380554-236380554 on Build GRCh38
rs184817273 Chr.2: 236380572-236380572 on Build GRCh38
rs373732714 Chr.2: 236380606-236380608 on Build GRCh38
rs919237316 Chr.2: 236380538-236380538 on Build GRCh38
rs562707115 Chr.2: 236380618-236380618 on Build GRCh38
rs189174983 Chr.2: 236380649-236380649 on Build GRCh38
rs997909321 Chr.2: 236380691-236380691 on Build GRCh38

View Full Product Details