Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47274330-47274601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB3
Entrez Gene ID
3690
Gene Name
integrin subunit beta 3
Gene Aliases
BDPLT16, BDPLT2, CD61, GP3A, GPIIIa, GT
UniGene
Hs.218040
Gene Chromosome Location
Chr.17: 47253842-47312711 on Build GRCh38
Transcripts
NM_000212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2696319 c.93T>C p.C31C Chr.17: 47274432-47274432 on Build GRCh38
6904890 c.80G>A p.G27E Chr.17: 47274419-47274419 on Build GRCh38
4268116 c.93T>C p.C31C Chr.17: 47274432-47274432 on Build GRCh38
6880382 c.10T>C p.W4R Chr.17: 47274490-47274490 on Build GRCh38
6904889 c.80G>A p.G27E Chr.17: 47274419-47274419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530929045 Chr.17: 47274488-47274488 on Build GRCh38
rs1024534966 Chr.17: 47274396-47274396 on Build GRCh38
rs1044985935 Chr.17: 47274526-47274526 on Build GRCh38
rs774170781 Chr.17: 47274525-47274525 on Build GRCh38
rs749206624 Chr.17: 47274515-47274515 on Build GRCh38
rs746091831 Chr.17: 47274495-47274495 on Build GRCh38
rs955185698 Chr.17: 47274469-47274469 on Build GRCh38
rs368427993 Chr.17: 47274461-47274461 on Build GRCh38
rs61736877 Chr.17: 47274435-47274435 on Build GRCh38
rs753146344 Chr.17: 47274450-47274450 on Build GRCh38
rs75427428 Chr.17: 47274439-47274439 on Build GRCh38
rs376323808 Chr.17: 47274546-47274546 on Build GRCh38
rs765882558 Chr.17: 47274440-47274440 on Build GRCh38
rs184077690 Chr.17: 47274571-47274571 on Build GRCh38
rs749837093 Chr.17: 47274548-47274548 on Build GRCh38
rs371555727 Chr.17: 47274486-47274486 on Build GRCh38
rs199802776 Chr.17: 47274384-47274384 on Build GRCh38
rs747375836 Chr.17: 47274481-47274481 on Build GRCh38
rs767197231 Chr.17: 47274537-47274537 on Build GRCh38
rs888066695 Chr.17: 47274499-47274499 on Build GRCh38
rs967611155 Chr.17: 47274424-47274424 on Build GRCh38
rs769937023 Chr.17: 47274510-47274510 on Build GRCh38
rs758852422 Chr.17: 47274459-47274459 on Build GRCh38
rs765826973 Chr.17: 47274549-47274549 on Build GRCh38
rs753378137 Chr.17: 47274556-47274556 on Build GRCh38
rs774042202 Chr.17: 47274382-47274382 on Build GRCh38
rs781627407 Chr.17: 47274494-47274494 on Build GRCh38
rs768512531 Chr.17: 47274524-47274524 on Build GRCh38
rs1013159150 Chr.17: 47274520-47274520 on Build GRCh38
rs1049301396 Chr.17: 47274465-47274465 on Build GRCh38
rs1014447426 Chr.17: 47274393-47274393 on Build GRCh38
rs544276300 Chr.17: 47274437-47274437 on Build GRCh38
rs775730681 Chr.17: 47274513-47274513 on Build GRCh38
rs761574328 Chr.17: 47274532-47274532 on Build GRCh38
rs772788858 Chr.17: 47274540-47274540 on Build GRCh38

View Full Product Details