Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 65420564-65420801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LEPR
Entrez Gene ID
3953
Gene Name
leptin receptor
Gene Aliases
CD295, LEP-R, LEPRD, OB-R, OBR
UniGene
Hs.23581
Gene Chromosome Location
Chr.1: 65420652-65637493 on Build GRCh38
Transcripts
NM_001003679.3 NM_001003680.3 NM_002303.5 NM_001198681.1 NM_001198683.1 NM_017526.4

Gene Symbol
LEPROT
Entrez Gene ID
54741
Gene Name
leptin receptor overlapping transcript
Gene Aliases
LEPR, OB-RGRP, OBRGRP, VPS55
UniGene
Hs.23581
Gene Chromosome Location
Chr.1: 65420448-65436007 on Build GRCh38
Transcripts
NM_001003679.3 NM_001003680.3 NM_002303.5 NM_001198681.1 NM_001198683.1 NM_017526.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7374931 c.15A>T p.K5N Chr.1: 65420739-65420739 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764647026 Chr.1: 65420738-65420738 on Build GRCh38
rs1049669496 Chr.1: 65420673-65420673 on Build GRCh38
rs932032488 Chr.1: 65420585-65420585 on Build GRCh38
rs763273103 Chr.1: 65420730-65420730 on Build GRCh38
rs776078871 Chr.1: 65420780-65420780 on Build GRCh38
rs930880776 Chr.1: 65420724-65420724 on Build GRCh38
rs770359733 Chr.1: 65420712-65420712 on Build GRCh38
rs3790434 Chr.1: 65420765-65420765 on Build GRCh38
rs752047665 Chr.1: 65420750-65420750 on Build GRCh38
rs770876789 Chr.1: 65420770-65420770 on Build GRCh38
rs898044832 Chr.1: 65420643-65420643 on Build GRCh38
rs778179157 Chr.1: 65420677-65420677 on Build GRCh38
rs767660276 Chr.1: 65420752-65420752 on Build GRCh38
rs955752657 Chr.1: 65420759-65420759 on Build GRCh38
rs575197913 Chr.1: 65420658-65420658 on Build GRCh38
rs371914392 Chr.1: 65420782-65420782 on Build GRCh38
rs189950527 Chr.1: 65420717-65420717 on Build GRCh38
rs775889131 Chr.1: 65420726-65420726 on Build GRCh38
rs565976952 Chr.1: 65420631-65420631 on Build GRCh38
rs760171463 Chr.1: 65420711-65420711 on Build GRCh38
rs1056216892 Chr.1: 65420729-65420729 on Build GRCh38
rs751663879 Chr.1: 65420678-65420678 on Build GRCh38
rs142626675 Chr.1: 65420635-65420635 on Build GRCh38
rs776131681 Chr.1: 65420714-65420714 on Build GRCh38
rs770940105 Chr.1: 65420776-65420776 on Build GRCh38
rs996932050 Chr.1: 65420653-65420653 on Build GRCh38
rs1050480676 Chr.1: 65420594-65420594 on Build GRCh38
rs968562467 Chr.1: 65420605-65420605 on Build GRCh38
rs778733406 Chr.1: 65420705-65420705 on Build GRCh38
rs998678922 Chr.1: 65420632-65420632 on Build GRCh38
rs372078226 Chr.1: 65420676-65420676 on Build GRCh38
rs1036606163 Chr.1: 65420640-65420640 on Build GRCh38
rs539912626 Chr.1: 65420762-65420762 on Build GRCh38
rs908292788 Chr.1: 65420684-65420684 on Build GRCh38
rs773083309 Chr.1: 65420708-65420708 on Build GRCh38
rs3790435 Chr.1: 65420715-65420715 on Build GRCh38
rs747933871 Chr.1: 65420706-65420706 on Build GRCh38
rs747963440 Chr.1: 65420763-65420763 on Build GRCh38
rs957262027 Chr.1: 65420749-65420749 on Build GRCh38
rs781428072 Chr.1: 65420688-65420688 on Build GRCh38
rs978978378 Chr.1: 65420636-65420636 on Build GRCh38
rs746032514 Chr.1: 65420692-65420692 on Build GRCh38
rs368145929 Chr.1: 65420675-65420675 on Build GRCh38
rs772024570 Chr.1: 65420707-65420707 on Build GRCh38
rs757467646 Chr.1: 65420681-65420681 on Build GRCh38
rs995016119 Chr.1: 65420672-65420672 on Build GRCh38
rs113211950 Chr.1: 65420769-65420769 on Build GRCh38
rs769934654 Chr.1: 65420702-65420702 on Build GRCh38
rs750566931 Chr.1: 65420761-65420761 on Build GRCh38
rs530298155 Chr.1: 65420784-65420784 on Build GRCh38

View Full Product Details