Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 21473443-21473670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NBPF3
Entrez Gene ID
84224
Gene Name
neuroblastoma breakpoint family member 3
Gene Aliases
AE2
UniGene
Hs.325422
Gene Chromosome Location
Chr.1: 21436775-21484900 on Build GRCh38
Transcripts
NM_001256416.2 NM_001256417.2 NM_032264.4 NR_046176.2 XM_006710957.1 XM_006710958.1 XM_006710959.2 XM_011542276.1 XM_011542277.1 XM_011542278.1 XM_011542281.1 XM_017002498.1 XM_017002499.1 XM_017002500.1 XM_017002501.1 XM_017002502.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5645555 c.803C>G p.S268* Chr.1: 21473448-21473448 on Build GRCh38
4606036 c.865G>A p.E289K Chr.1: 21473510-21473510 on Build GRCh38
4028302 c.922G>A p.A308T Chr.1: 21473567-21473567 on Build GRCh38
6009136 c.816C>T p.H272H Chr.1: 21473461-21473461 on Build GRCh38
6426659 c.857C>A p.T286K Chr.1: 21473502-21473502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762367983 Chr.1: 21473588-21473588 on Build GRCh38
rs774129980 Chr.1: 21473550-21473550 on Build GRCh38
rs780717262 Chr.1: 21473521-21473521 on Build GRCh38
rs770888853 Chr.1: 21473544-21473544 on Build GRCh38
rs910122861 Chr.1: 21473537-21473537 on Build GRCh38
rs993028908 Chr.1: 21473477-21473477 on Build GRCh38
rs764317550 Chr.1: 21473585-21473585 on Build GRCh38
rs983008175 Chr.1: 21473514-21473515 on Build GRCh38
rs755745119 Chr.1: 21473588-21473588 on Build GRCh38
rs202196655 Chr.1: 21473559-21473559 on Build GRCh38
rs747693353 Chr.1: 21473525-21473525 on Build GRCh38
rs757130243 Chr.1: 21473626-21473626 on Build GRCh38
rs767170688 Chr.1: 21473603-21473603 on Build GRCh38
rs750831912 Chr.1: 21473482-21473482 on Build GRCh38
rs769337272 Chr.1: 21473527-21473527 on Build GRCh38
rs149171985 Chr.1: 21473475-21473475 on Build GRCh38
rs752765705 Chr.1: 21473466-21473466 on Build GRCh38
rs752344449 Chr.1: 21473607-21473607 on Build GRCh38
rs200424283 Chr.1: 21473514-21473514 on Build GRCh38
rs779369557 Chr.1: 21473508-21473508 on Build GRCh38
rs573244682 Chr.1: 21473611-21473611 on Build GRCh38
rs868120491 Chr.1: 21473563-21473564 on Build GRCh38
rs765841678 Chr.1: 21473595-21473595 on Build GRCh38
rs757672572 Chr.1: 21473507-21473507 on Build GRCh38
rs750737912 Chr.1: 21473600-21473600 on Build GRCh38
rs749177270 Chr.1: 21473534-21473534 on Build GRCh38
rs376234807 Chr.1: 21473622-21473622 on Build GRCh38
rs772325096 Chr.1: 21473553-21473553 on Build GRCh38
rs373276794 Chr.1: 21473572-21473572 on Build GRCh38
rs777227431 Chr.1: 21473621-21473621 on Build GRCh38
rs138236134 Chr.1: 21473533-21473533 on Build GRCh38
rs143299774 Chr.1: 21473485-21473485 on Build GRCh38
rs758743882 Chr.1: 21473602-21473602 on Build GRCh38
rs760805949 Chr.1: 21473565-21473565 on Build GRCh38
rs200944200 Chr.1: 21473512-21473512 on Build GRCh38
rs747645859 Chr.1: 21473564-21473565 on Build GRCh38
rs141340113 Chr.1: 21473486-21473486 on Build GRCh38
rs941715526 Chr.1: 21473562-21473562 on Build GRCh38
rs759263409 Chr.1: 21473552-21473552 on Build GRCh38
rs753986881 Chr.1: 21473586-21473586 on Build GRCh38
rs750781298 Chr.1: 21473510-21473510 on Build GRCh38

View Full Product Details