Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153626261-153626522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S100A13
Entrez Gene ID
6284
Gene Name
S100 calcium binding protein A13
Gene Aliases
-
UniGene
Hs.516505
Gene Chromosome Location
Chr.1: 153618799-153634092 on Build GRCh38
Transcripts
NM_001024210.1 NM_001024211.1 NM_001024212.1 NM_001024213.1 NM_005979.2 XM_005245434.3 XM_011509862.2 XM_011509863.2 XM_011509864.1 XM_017002034.1 XM_017002035.1 XM_017002036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6249270 c.108C>A p.N36K Chr.1: 153626365-153626365 on Build GRCh38
7145629 c.2T>G p.M1R Chr.1: 153626471-153626471 on Build GRCh38
6056082 c.86G>C p.R29P Chr.1: 153626387-153626387 on Build GRCh38
206329 c.86G>A p.R29Q Chr.1: 153626387-153626387 on Build GRCh38
6771198 c.108C>T p.N36N Chr.1: 153626365-153626365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772347756 Chr.1: 153626369-153626369 on Build GRCh38
rs758012417 Chr.1: 153626289-153626289 on Build GRCh38
rs1026364033 Chr.1: 153626300-153626300 on Build GRCh38
rs779000547 Chr.1: 153626458-153626458 on Build GRCh38
rs377108150 Chr.1: 153626357-153626357 on Build GRCh38
rs199830698 Chr.1: 153626366-153626366 on Build GRCh38
rs754572136 Chr.1: 153626310-153626310 on Build GRCh38
rs778292793 Chr.1: 153626433-153626433 on Build GRCh38
rs372709618 Chr.1: 153626493-153626493 on Build GRCh38
rs749592486 Chr.1: 153626452-153626452 on Build GRCh38
rs139231458 Chr.1: 153626393-153626393 on Build GRCh38
rs202195411 Chr.1: 153626330-153626330 on Build GRCh38
rs778131659 Chr.1: 153626337-153626337 on Build GRCh38
rs374018193 Chr.1: 153626332-153626332 on Build GRCh38
rs1007313349 Chr.1: 153626440-153626440 on Build GRCh38
rs765079870 Chr.1: 153626371-153626371 on Build GRCh38
rs770083487 Chr.1: 153626331-153626331 on Build GRCh38
rs879076165 Chr.1: 153626389-153626389 on Build GRCh38
rs199790715 Chr.1: 153626427-153626427 on Build GRCh38
rs369833765 Chr.1: 153626364-153626364 on Build GRCh38
rs373852522 Chr.1: 153626370-153626370 on Build GRCh38
rs993364090 Chr.1: 153626403-153626403 on Build GRCh38
rs754377096 Chr.1: 153626422-153626422 on Build GRCh38
rs770595516 Chr.1: 153626457-153626457 on Build GRCh38
rs746107807 Chr.1: 153626465-153626465 on Build GRCh38
rs751005885 Chr.1: 153626307-153626307 on Build GRCh38
rs750278910 Chr.1: 153626382-153626382 on Build GRCh38
rs781660855 Chr.1: 153626323-153626323 on Build GRCh38
rs4879 Chr.1: 153626431-153626431 on Build GRCh38
rs762849446 Chr.1: 153626388-153626388 on Build GRCh38
rs765895178 Chr.1: 153626480-153626481 on Build GRCh38
rs769828455 Chr.1: 153626501-153626501 on Build GRCh38
rs771026346 Chr.1: 153626362-153626362 on Build GRCh38
rs751290059 Chr.1: 153626412-153626412 on Build GRCh38
rs530505141 Chr.1: 153626468-153626468 on Build GRCh38
rs778915425 Chr.1: 153626335-153626335 on Build GRCh38
rs779243655 Chr.1: 153626365-153626365 on Build GRCh38
rs11388 Chr.1: 153626386-153626386 on Build GRCh38
rs565398385 Chr.1: 153626305-153626305 on Build GRCh38
rs113541380 Chr.1: 153626428-153626428 on Build GRCh38
rs370377697 Chr.1: 153626390-153626390 on Build GRCh38
rs775734935 Chr.1: 153626473-153626473 on Build GRCh38

View Full Product Details