Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 16326632-16326873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXNAD1
Entrez Gene ID
92106
Gene Name
oxidoreductase NAD binding domain containing 1
Gene Aliases
-
UniGene
Hs.655449
Gene Chromosome Location
Chr.3: 16265160-16386979 on Build GRCh38
Transcripts
XM_011534232.2 XM_011534233.2 XM_011534234.2 XM_017007485.1 XM_017007488.1 XM_017007489.1 XM_017007491.1 XM_017007494.1 XM_017007496.1 NM_015150.1 XM_005264985.1 XM_005264986.2 XM_006713069.1 XM_011533529.1 XM_011533530.1 XM_011533531.1 XM_017006005.1 XM_017006006.1 XM_017006007.1

Gene Symbol
RFTN1
Entrez Gene ID
23180
Gene Name
raftlin, lipid raft linker 1
Gene Aliases
MIG2, PIB10, PIG9, RAFTLIN
UniGene
Hs.98910
Gene Chromosome Location
Chr.3: 16315845-16513715 on Build GRCh38
Transcripts
XM_011534232.2 XM_011534233.2 XM_011534234.2 XM_017007485.1 XM_017007488.1 XM_017007489.1 XM_017007491.1 XM_017007494.1 XM_017007496.1 NM_015150.1 XM_005264985.1 XM_005264986.2 XM_006713069.1 XM_011533529.1 XM_011533530.1 XM_011533531.1 XM_017006005.1 XM_017006006.1 XM_017006007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1752996 c.1177C>T p.L393L Chr.3: 16326846-16326846 on Build GRCh38
479652 c.1199A>G p.Y400C Chr.3: 16326824-16326824 on Build GRCh38
3589530 c.1184A>G p.N395S Chr.3: 16326839-16326839 on Build GRCh38
291619 c.1167C>G p.D389E Chr.3: 16326856-16326856 on Build GRCh38
4584079 c.1233C>T p.P411P Chr.3: 16326790-16326790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046629163 Chr.3: 16326700-16326700 on Build GRCh38
rs923948345 Chr.3: 16326655-16326655 on Build GRCh38
rs776367773 Chr.3: 16326836-16326836 on Build GRCh38
rs954950008 Chr.3: 16326788-16326788 on Build GRCh38
rs866617003 Chr.3: 16326748-16326748 on Build GRCh38
rs916876318 Chr.3: 16326689-16326689 on Build GRCh38
rs760229079 Chr.3: 16326829-16326829 on Build GRCh38
rs763788878 Chr.3: 16326742-16326742 on Build GRCh38
rs138362770 Chr.3: 16326768-16326768 on Build GRCh38
rs958028639 Chr.3: 16326666-16326666 on Build GRCh38
rs755067050 Chr.3: 16326781-16326781 on Build GRCh38
rs149250933 Chr.3: 16326789-16326789 on Build GRCh38
rs368117034 Chr.3: 16326766-16326766 on Build GRCh38
rs748372599 Chr.3: 16326786-16326786 on Build GRCh38
rs772014458 Chr.3: 16326802-16326802 on Build GRCh38
rs768360426 Chr.3: 16326675-16326675 on Build GRCh38
rs910629121 Chr.3: 16326832-16326832 on Build GRCh38
rs374908747 Chr.3: 16326730-16326730 on Build GRCh38
rs377108354 Chr.3: 16326835-16326835 on Build GRCh38
rs750914402 Chr.3: 16326767-16326767 on Build GRCh38
rs756839113 Chr.3: 16326766-16326767 on Build GRCh38
rs998047684 Chr.3: 16326824-16326824 on Build GRCh38
rs747085173 Chr.3: 16326780-16326780 on Build GRCh38
rs780469915 Chr.3: 16326770-16326770 on Build GRCh38
rs753612661 Chr.3: 16326743-16326743 on Build GRCh38
rs745751576 Chr.3: 16326799-16326799 on Build GRCh38
rs544519382 Chr.3: 16326696-16326696 on Build GRCh38
rs761582997 Chr.3: 16326841-16326841 on Build GRCh38
rs572578132 Chr.3: 16326695-16326695 on Build GRCh38
rs978121093 Chr.3: 16326654-16326654 on Build GRCh38
rs928100236 Chr.3: 16326707-16326707 on Build GRCh38
rs1031548243 Chr.3: 16326773-16326773 on Build GRCh38
rs541489584 Chr.3: 16326737-16326737 on Build GRCh38
rs781385817 Chr.3: 16326784-16326784 on Build GRCh38
rs1013016254 Chr.3: 16326676-16326676 on Build GRCh38
rs757629774 Chr.3: 16326753-16326753 on Build GRCh38
rs775348055 Chr.3: 16326816-16326816 on Build GRCh38
rs139132308 Chr.3: 16326790-16326790 on Build GRCh38
rs1021524167 Chr.3: 16326690-16326690 on Build GRCh38

View Full Product Details