Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43139165-43139424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTK7
Entrez Gene ID
5754
Gene Name
protein tyrosine kinase 7 (inactive)
Gene Aliases
CCK-4, CCK4
UniGene
Hs.90572
Gene Chromosome Location
Chr.6: 43076268-43161720 on Build GRCh38
Transcripts
NM_001270398.1 NM_002821.4 NM_152880.3 NM_152881.3 NM_152882.3 NR_072997.1 NR_072998.1 XM_011514765.2 XM_011514766.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1079458 c.1410C>T p.S470S Chr.6: 43139183-43139183 on Build GRCh38
1643078 c.1470C>T p.I490I Chr.6: 43139243-43139243 on Build GRCh38
5737605 c.776G>A p.R259H Chr.6: 43139212-43139212 on Build GRCh38
3079828 c.1400G>A p.R467H Chr.6: 43139173-43139173 on Build GRCh38
1444647 c.1480G>A p.A494T Chr.6: 43139253-43139253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769889457 Chr.6: 43139373-43139373 on Build GRCh38
rs752545639 Chr.6: 43139184-43139184 on Build GRCh38
rs369849121 Chr.6: 43139290-43139290 on Build GRCh38
rs199696954 Chr.6: 43139351-43139351 on Build GRCh38
rs778092321 Chr.6: 43139246-43139246 on Build GRCh38
rs765803186 Chr.6: 43139358-43139358 on Build GRCh38
rs759377536 Chr.6: 43139257-43139257 on Build GRCh38
rs372585693 Chr.6: 43139234-43139234 on Build GRCh38
rs746019072 Chr.6: 43139243-43139243 on Build GRCh38
rs941282914 Chr.6: 43139279-43139279 on Build GRCh38
rs753279775 Chr.6: 43139361-43139361 on Build GRCh38
rs918409812 Chr.6: 43139339-43139339 on Build GRCh38
rs780428018 Chr.6: 43139375-43139377 on Build GRCh38
rs149972636 Chr.6: 43139239-43139239 on Build GRCh38
rs776835676 Chr.6: 43139255-43139255 on Build GRCh38
rs781109466 Chr.6: 43139368-43139368 on Build GRCh38
rs746000560 Chr.6: 43139370-43139370 on Build GRCh38
rs974008236 Chr.6: 43139289-43139289 on Build GRCh38
rs763772683 Chr.6: 43139204-43139204 on Build GRCh38
rs772596018 Chr.6: 43139245-43139245 on Build GRCh38
rs966560025 Chr.6: 43139252-43139252 on Build GRCh38
rs754299400 Chr.6: 43139227-43139227 on Build GRCh38
rs775234118 Chr.6: 43139271-43139271 on Build GRCh38
rs61996307 Chr.6: 43139224-43139224 on Build GRCh38
rs201529711 Chr.6: 43139250-43139250 on Build GRCh38
rs373305442 Chr.6: 43139362-43139362 on Build GRCh38
rs756405228 Chr.6: 43139318-43139327 on Build GRCh38
rs1039700470 Chr.6: 43139376-43139376 on Build GRCh38
rs761529065 Chr.6: 43139307-43139307 on Build GRCh38
rs936125528 Chr.6: 43139344-43139344 on Build GRCh38
rs750001479 Chr.6: 43139320-43139320 on Build GRCh38
rs925108741 Chr.6: 43139338-43139338 on Build GRCh38
rs757556798 Chr.6: 43139367-43139367 on Build GRCh38
rs749234249 Chr.6: 43139377-43139377 on Build GRCh38
rs571946445 Chr.6: 43139186-43139186 on Build GRCh38
rs762692851 Chr.6: 43139282-43139282 on Build GRCh38
rs780764712 Chr.6: 43139226-43139226 on Build GRCh38
rs201674266 Chr.6: 43139183-43139183 on Build GRCh38
rs545149626 Chr.6: 43139392-43139392 on Build GRCh38
rs373421990 Chr.6: 43139393-43139393 on Build GRCh38
rs377486112 Chr.6: 43139363-43139363 on Build GRCh38
rs775353315 Chr.6: 43139374-43139374 on Build GRCh38
rs747269276 Chr.6: 43139248-43139248 on Build GRCh38
rs929767656 Chr.6: 43139350-43139350 on Build GRCh38
rs199978430 Chr.6: 43139284-43139284 on Build GRCh38
rs748567987 Chr.6: 43139244-43139244 on Build GRCh38
rs534553772 Chr.6: 43139192-43139192 on Build GRCh38
rs755412410 Chr.6: 43139356-43139356 on Build GRCh38
rs950091438 Chr.6: 43139354-43139354 on Build GRCh38
rs369502357 Chr.6: 43139382-43139382 on Build GRCh38
rs145195593 Chr.6: 43139249-43139249 on Build GRCh38
rs144938642 Chr.6: 43139212-43139212 on Build GRCh38
rs369263649 Chr.6: 43139217-43139217 on Build GRCh38
rs766925779 Chr.6: 43139315-43139315 on Build GRCh38

View Full Product Details